Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 106903072-106903339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN2
Entrez Gene ID
9075
Gene Name
claudin 2
Gene Aliases
-
UniGene
Hs.522746
Gene Chromosome Location
Chr.X: 106900164-106930861 on Build GRCh38
Transcripts
NM_001171092.1 NM_001171706.1 NM_138382.2

Gene Symbol
RIPPLY1
Entrez Gene ID
92129
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
UniGene
Hs.334726
Gene Chromosome Location
Chr.X: 106900063-106903331 on Build GRCh38
Transcripts
NM_001171092.1 NM_001171706.1 NM_138382.2

SNP Detail

SNP ID Chromosome Location
rs768305648 Chr.X: 106903153-106903153 on Build GRCh38
rs367555601 Chr.X: 106903253-106903253 on Build GRCh38
rs968459718 Chr.X: 106903231-106903231 on Build GRCh38
rs779551049 Chr.X: 106903141-106903141 on Build GRCh38
rs371828879 Chr.X: 106903281-106903281 on Build GRCh38
rs747842759 Chr.X: 106903178-106903178 on Build GRCh38
rs757086100 Chr.X: 106903106-106903106 on Build GRCh38
rs138679347 Chr.X: 106903288-106903288 on Build GRCh38
rs778515799 Chr.X: 106903172-106903172 on Build GRCh38
rs765447238 Chr.X: 106903261-106903261 on Build GRCh38
rs771369948 Chr.X: 106903205-106903205 on Build GRCh38
rs992310080 Chr.X: 106903293-106903293 on Build GRCh38
rs766249767 Chr.X: 106903294-106903294 on Build GRCh38
rs748782133 Chr.X: 106903149-106903149 on Build GRCh38
rs201845535 Chr.X: 106903197-106903197 on Build GRCh38
rs371861405 Chr.X: 106903292-106903292 on Build GRCh38
rs200086437 Chr.X: 106903132-106903132 on Build GRCh38
rs753864349 Chr.X: 106903314-106903314 on Build GRCh38
rs756017902 Chr.X: 106903133-106903133 on Build GRCh38
rs1030062506 Chr.X: 106903188-106903188 on Build GRCh38
rs775849540 Chr.X: 106903271-106903271 on Build GRCh38
rs202113992 Chr.X: 106903255-106903255 on Build GRCh38
rs374926749 Chr.X: 106903287-106903287 on Build GRCh38
rs765582615 Chr.X: 106903259-106903261 on Build GRCh38
rs772423682 Chr.X: 106903189-106903189 on Build GRCh38
rs5962774 Chr.X: 106903119-106903119 on Build GRCh38
rs753793833 Chr.X: 106903123-106903123 on Build GRCh38
rs375592382 Chr.X: 106903194-106903194 on Build GRCh38

View Full Product Details