Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 81174735-81174988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
UniGene
Hs.259559
Gene Chromosome Location
Chr.12: 81078018-81257795 on Build GRCh38
Transcripts
NM_024560.2 XM_005269150.2 XM_005269151.4 XM_011538740.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5630487 c.1123G>A p.A375T Chr.12: 81174812-81174812 on Build GRCh38
3465887 c.1201C>T p.R401C Chr.12: 81174890-81174890 on Build GRCh38
6138438 c.1217G>T p.G406V Chr.12: 81174906-81174906 on Build GRCh38
1299902 c.1186G>A p.A396T Chr.12: 81174875-81174875 on Build GRCh38
231701 c.1124C>T p.A375V Chr.12: 81174813-81174813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369700597 Chr.12: 81174774-81174774 on Build GRCh38
rs185881304 Chr.12: 81174890-81174890 on Build GRCh38
rs574304962 Chr.12: 81174959-81174959 on Build GRCh38
rs753698444 Chr.12: 81174773-81174773 on Build GRCh38
rs138734052 Chr.12: 81174820-81174820 on Build GRCh38
rs754483699 Chr.12: 81174891-81174891 on Build GRCh38
rs937541558 Chr.12: 81174785-81174785 on Build GRCh38
rs200388083 Chr.12: 81174779-81174779 on Build GRCh38
rs747340180 Chr.12: 81174818-81174818 on Build GRCh38
rs752250118 Chr.12: 81174902-81174902 on Build GRCh38
rs555708909 Chr.12: 81174917-81174917 on Build GRCh38
rs766419739 Chr.12: 81174888-81174888 on Build GRCh38
rs748539356 Chr.12: 81174804-81174804 on Build GRCh38
rs902243280 Chr.12: 81174872-81174872 on Build GRCh38
rs756520749 Chr.12: 81174905-81174905 on Build GRCh38
rs761700970 Chr.12: 81174846-81174846 on Build GRCh38
rs762839597 Chr.12: 81174887-81174887 on Build GRCh38
rs754747641 Chr.12: 81174787-81174787 on Build GRCh38
rs141866758 Chr.12: 81174892-81174892 on Build GRCh38
rs1019502101 Chr.12: 81174912-81174912 on Build GRCh38
rs201458439 Chr.12: 81174845-81174845 on Build GRCh38
rs573499661 Chr.12: 81174874-81174874 on Build GRCh38
rs767694781 Chr.12: 81174770-81174770 on Build GRCh38
rs765226724 Chr.12: 81174778-81174778 on Build GRCh38
rs778371758 Chr.12: 81174818-81174818 on Build GRCh38
rs146040643 Chr.12: 81174812-81174812 on Build GRCh38
rs953889155 Chr.12: 81174899-81174899 on Build GRCh38
rs763643365 Chr.12: 81174843-81174843 on Build GRCh38
rs199939705 Chr.12: 81174907-81174907 on Build GRCh38
rs768892088 Chr.12: 81174945-81174945 on Build GRCh38
rs374493855 Chr.12: 81174956-81174956 on Build GRCh38
rs757770927 Chr.12: 81174914-81174914 on Build GRCh38
rs772337973 Chr.12: 81174841-81174841 on Build GRCh38
rs771442183 Chr.12: 81174827-81174827 on Build GRCh38
rs774453986 Chr.12: 81174828-81174828 on Build GRCh38
rs377119299 Chr.12: 81174798-81174798 on Build GRCh38
rs759607208 Chr.12: 81174832-81174832 on Build GRCh38
rs375680193 Chr.12: 81174850-81174850 on Build GRCh38
rs751086812 Chr.12: 81174780-81174780 on Build GRCh38
rs745958355 Chr.12: 81174922-81174922 on Build GRCh38
rs775712918 Chr.12: 81174944-81174944 on Build GRCh38

View Full Product Details