Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95613763-95614026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
UniGene
Hs.500645
Gene Chromosome Location
Chr.10: 95605929-95656810 on Build GRCh38
Transcripts
NM_001017423.1 NM_001323412.1 NM_001323413.1 NM_001323414.1 NM_001323415.1 NM_001323416.1 NM_001323417.1 NM_001323418.1 NM_001323419.1 NM_002860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836327 c.1783G>C p.D595H Chr.10: 95613984-95613984 on Build GRCh38
6210730 c.1809C>T p.D603D Chr.10: 95613856-95613856 on Build GRCh38
5629956 c.1797G>C p.R599S Chr.10: 95613970-95613970 on Build GRCh38
6598157 c.1756A>G p.M586V Chr.10: 95614011-95614011 on Build GRCh38
6598149 c.1889T>G p.F630C Chr.10: 95613776-95613776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760405893 Chr.10: 95613956-95613956 on Build GRCh38
rs577073953 Chr.10: 95613944-95613944 on Build GRCh38
rs528157362 Chr.10: 95613974-95613974 on Build GRCh38
rs11541780 Chr.10: 95613997-95613997 on Build GRCh38
rs369452137 Chr.10: 95613903-95613903 on Build GRCh38
rs771009824 Chr.10: 95613935-95613935 on Build GRCh38
rs748295825 Chr.10: 95613912-95613912 on Build GRCh38
rs372599895 Chr.10: 95613978-95613978 on Build GRCh38
rs753352541 Chr.10: 95613866-95613866 on Build GRCh38
rs752764884 Chr.10: 95613970-95613970 on Build GRCh38
rs373988688 Chr.10: 95613916-95613916 on Build GRCh38
rs997635391 Chr.10: 95613937-95613937 on Build GRCh38
rs200630216 Chr.10: 95613976-95613976 on Build GRCh38
rs200514228 Chr.10: 95613936-95613936 on Build GRCh38
rs574468427 Chr.10: 95613854-95613854 on Build GRCh38
rs774274057 Chr.10: 95613800-95613800 on Build GRCh38
rs371794461 Chr.10: 95613884-95613884 on Build GRCh38
rs770815414 Chr.10: 95613798-95613798 on Build GRCh38
rs545630796 Chr.10: 95613948-95613948 on Build GRCh38
rs777887374 Chr.10: 95613922-95613922 on Build GRCh38
rs543845150 Chr.10: 95613860-95613860 on Build GRCh38
rs41291564 Chr.10: 95613987-95613987 on Build GRCh38
rs752173917 Chr.10: 95613900-95613900 on Build GRCh38
rs376515375 Chr.10: 95613896-95613896 on Build GRCh38
rs766756427 Chr.10: 95613885-95613886 on Build GRCh38
rs754097296 Chr.10: 95613887-95613900 on Build GRCh38
rs755594128 Chr.10: 95613996-95613996 on Build GRCh38
rs764910330 Chr.10: 95613979-95613979 on Build GRCh38
rs759267778 Chr.10: 95613801-95613801 on Build GRCh38
rs141192935 Chr.10: 95613967-95613967 on Build GRCh38
rs201126424 Chr.10: 95613934-95613934 on Build GRCh38
rs775400551 Chr.10: 95613952-95613952 on Build GRCh38
rs113345547 Chr.10: 95613917-95613917 on Build GRCh38
rs756836950 Chr.10: 95613890-95613890 on Build GRCh38
rs767355400 Chr.10: 95613830-95613830 on Build GRCh38
rs759835055 Chr.10: 95613906-95613906 on Build GRCh38
rs752285111 Chr.10: 95613838-95613838 on Build GRCh38

View Full Product Details