Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68018546-68018814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3B1
Entrez Gene ID
221
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
ALDH4, ALDH7
UniGene
Hs.523841
Gene Chromosome Location
Chr.11: 68008547-68029282 on Build GRCh38
Transcripts
NM_000694.3 NM_001030010.2 NM_001161473.2 NM_001290058.1 NM_001290059.1

SNP Detail

SNP ID Chromosome Location
rs747052509 Chr.11: 68018662-68018662 on Build GRCh38
rs766312412 Chr.11: 68018698-68018698 on Build GRCh38
rs1044126278 Chr.11: 68018666-68018666 on Build GRCh38
rs759805249 Chr.11: 68018730-68018730 on Build GRCh38
rs1009989026 Chr.11: 68018719-68018719 on Build GRCh38
rs777394092 Chr.11: 68018578-68018578 on Build GRCh38
rs771001411 Chr.11: 68018665-68018665 on Build GRCh38
rs773150926 Chr.11: 68018659-68018659 on Build GRCh38
rs906456630 Chr.11: 68018739-68018739 on Build GRCh38
rs376553943 Chr.11: 68018778-68018778 on Build GRCh38
rs959096514 Chr.11: 68018598-68018598 on Build GRCh38
rs943422808 Chr.11: 68018686-68018686 on Build GRCh38
rs780278979 Chr.11: 68018593-68018594 on Build GRCh38
rs773401320 Chr.11: 68018646-68018646 on Build GRCh38
rs397777471 Chr.11: 68018596-68018597 on Build GRCh38
rs764608175 Chr.11: 68018756-68018756 on Build GRCh38
rs745605228 Chr.11: 68018589-68018589 on Build GRCh38
rs775991118 Chr.11: 68018734-68018734 on Build GRCh38
rs752017086 Chr.11: 68018758-68018758 on Build GRCh38
rs766644337 Chr.11: 68018604-68018604 on Build GRCh38
rs763357662 Chr.11: 68018744-68018744 on Build GRCh38
rs765673658 Chr.11: 68018565-68018565 on Build GRCh38
rs746611472 Chr.11: 68018580-68018580 on Build GRCh38
rs768701840 Chr.11: 68018658-68018658 on Build GRCh38
rs1017098544 Chr.11: 68018766-68018766 on Build GRCh38
rs750074611 Chr.11: 68018765-68018765 on Build GRCh38
rs776851485 Chr.11: 68018605-68018605 on Build GRCh38
rs1051933826 Chr.11: 68018649-68018649 on Build GRCh38
rs199923261 Chr.11: 68018594-68018594 on Build GRCh38
rs868425748 Chr.11: 68018597-68018598 on Build GRCh38
rs758986244 Chr.11: 68018618-68018618 on Build GRCh38
rs757799681 Chr.11: 68018572-68018572 on Build GRCh38
rs760799429 Chr.11: 68018601-68018601 on Build GRCh38
rs756735295 Chr.11: 68018640-68018640 on Build GRCh38
rs767161715 Chr.11: 68018760-68018760 on Build GRCh38
rs961037941 Chr.11: 68018771-68018771 on Build GRCh38
rs529954694 Chr.11: 68018591-68018591 on Build GRCh38
rs770114586 Chr.11: 68018731-68018731 on Build GRCh38
rs368781207 Chr.11: 68018621-68018621 on Build GRCh38
rs781125264 Chr.11: 68018586-68018586 on Build GRCh38
rs961353800 Chr.11: 68018642-68018642 on Build GRCh38
rs377044666 Chr.11: 68018695-68018695 on Build GRCh38
rs34280013 Chr.11: 68018790-68018790 on Build GRCh38
rs1003321518 Chr.11: 68018582-68018582 on Build GRCh38
rs369048585 Chr.11: 68018654-68018654 on Build GRCh38
rs374693803 Chr.11: 68018648-68018648 on Build GRCh38
rs778864435 Chr.11: 68018755-68018755 on Build GRCh38
rs759827442 Chr.11: 68018609-68018609 on Build GRCh38
rs868314955 Chr.11: 68018575-68018575 on Build GRCh38
rs757002126 Chr.11: 68018584-68018584 on Build GRCh38
rs3815220 Chr.11: 68018644-68018644 on Build GRCh38
rs1054296261 Chr.11: 68018713-68018713 on Build GRCh38
rs570844706 Chr.11: 68018759-68018759 on Build GRCh38
rs373414669 Chr.11: 68018685-68018685 on Build GRCh38
rs755926658 Chr.11: 68018645-68018645 on Build GRCh38
rs1042423488 Chr.11: 68018728-68018728 on Build GRCh38
rs754887884 Chr.11: 68018626-68018626 on Build GRCh38
rs753181418 Chr.11: 68018616-68018616 on Build GRCh38
rs780929250 Chr.11: 68018643-68018643 on Build GRCh38
rs753205833 Chr.11: 68018569-68018569 on Build GRCh38
rs765620459 Chr.11: 68018613-68018613 on Build GRCh38
rs893767207 Chr.11: 68018714-68018714 on Build GRCh38
rs566880821 Chr.11: 68018635-68018635 on Build GRCh38
rs772020036 Chr.11: 68018599-68018599 on Build GRCh38
rs949765784 Chr.11: 68018663-68018663 on Build GRCh38
rs773376825 Chr.11: 68018600-68018600 on Build GRCh38

View Full Product Details