Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118196626-118196899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAML
Entrez Gene ID
120425
Gene Name
junction adhesion molecule like
Gene Aliases
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638
UniGene
Hs.16291
Gene Chromosome Location
Chr.11: 118193727-118225094 on Build GRCh38
Transcripts
NM_001098526.1 NM_001286570.1 NM_001286571.1 NM_153206.2 NR_104479.1 XM_005271407.3 XM_017017213.1 XM_017017214.1 XM_017017215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384095 c.1000G>T p.V334L Chr.11: 118196797-118196797 on Build GRCh38
4189224 c.1024_1025insA p.P343fs*3 Chr.11: 118196772-118196773 on Build GRCh38
3443892 c.1089C>T p.T363T Chr.11: 118196738-118196738 on Build GRCh38
7264440 c.1088C>T p.T363I Chr.11: 118196739-118196739 on Build GRCh38
6599215 c.1044C>T p.I348I Chr.11: 118196783-118196783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs648947 Chr.11: 118196711-118196711 on Build GRCh38
rs34170062 Chr.11: 118196689-118196689 on Build GRCh38
rs1038260157 Chr.11: 118196726-118196726 on Build GRCh38
rs967518194 Chr.11: 118196647-118196647 on Build GRCh38
rs777112471 Chr.11: 118196755-118196755 on Build GRCh38
rs138630785 Chr.11: 118196807-118196807 on Build GRCh38
rs139585653 Chr.11: 118196722-118196722 on Build GRCh38
rs763049485 Chr.11: 118196782-118196782 on Build GRCh38
rs753704215 Chr.11: 118196794-118196794 on Build GRCh38
rs757991355 Chr.11: 118196824-118196824 on Build GRCh38
rs755866037 Chr.11: 118196684-118196684 on Build GRCh38
rs753259263 Chr.11: 118196691-118196691 on Build GRCh38
rs576482997 Chr.11: 118196803-118196803 on Build GRCh38
rs749019316 Chr.11: 118196702-118196702 on Build GRCh38
rs746488835 Chr.11: 118196832-118196832 on Build GRCh38
rs770171107 Chr.11: 118196696-118196697 on Build GRCh38
rs1050807296 Chr.11: 118196756-118196756 on Build GRCh38
rs559030480 Chr.11: 118196875-118196875 on Build GRCh38
rs946507048 Chr.11: 118196654-118196654 on Build GRCh38
rs372067604 Chr.11: 118196841-118196841 on Build GRCh38
rs267602712 Chr.11: 118196764-118196764 on Build GRCh38
rs770102239 Chr.11: 118196868-118196868 on Build GRCh38
rs918437049 Chr.11: 118196744-118196744 on Build GRCh38
rs150082618 Chr.11: 118196779-118196779 on Build GRCh38
rs755282489 Chr.11: 118196788-118196788 on Build GRCh38
rs78109903 Chr.11: 118196766-118196766 on Build GRCh38
rs141143931 Chr.11: 118196822-118196822 on Build GRCh38
rs376459165 Chr.11: 118196710-118196710 on Build GRCh38
rs981529954 Chr.11: 118196659-118196659 on Build GRCh38
rs765775784 Chr.11: 118196774-118196774 on Build GRCh38
rs771661391 Chr.11: 118196715-118196715 on Build GRCh38
rs748773343 Chr.11: 118196673-118196681 on Build GRCh38
rs777565311 Chr.11: 118196688-118196688 on Build GRCh38
rs1029732298 Chr.11: 118196850-118196850 on Build GRCh38
rs754864453 Chr.11: 118196869-118196869 on Build GRCh38
rs764658369 Chr.11: 118196747-118196747 on Build GRCh38
rs776118584 Chr.11: 118196858-118196858 on Build GRCh38
rs370877046 Chr.11: 118196829-118196829 on Build GRCh38
rs186409060 Chr.11: 118196677-118196677 on Build GRCh38
rs955045444 Chr.11: 118196865-118196865 on Build GRCh38
rs958729883 Chr.11: 118196680-118196680 on Build GRCh38
rs748562904 Chr.11: 118196866-118196866 on Build GRCh38
rs775038870 Chr.11: 118196719-118196719 on Build GRCh38
rs766414433 Chr.11: 118196783-118196783 on Build GRCh38
rs757934856 Chr.11: 118196802-118196802 on Build GRCh38
rs979300955 Chr.11: 118196663-118196663 on Build GRCh38
rs928669418 Chr.11: 118196671-118196671 on Build GRCh38
rs749963177 Chr.11: 118196793-118196793 on Build GRCh38

View Full Product Details