Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34983211-34983464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1A
Entrez Gene ID
23294
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Gene Aliases
ANKS1
UniGene
Hs.656492
Gene Chromosome Location
Chr.6: 34889261-35097994 on Build GRCh38
Transcripts
NM_015245.2 XM_005248964.3 XM_006715036.2 XM_011514431.2 XM_011514432.2 XM_011514433.1 XM_011514434.2 XM_011514435.2 XM_011514436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405070 c.956C>T p.P319L Chr.6: 34983369-34983369 on Build GRCh38
5560421 c.985A>G p.M329V Chr.6: 34983398-34983398 on Build GRCh38
4856626 c.992C>G p.S331C Chr.6: 34983405-34983405 on Build GRCh38
5553261 c.940G>T p.E314* Chr.6: 34983353-34983353 on Build GRCh38
4562509 c.929G>A p.R310K Chr.6: 34983342-34983342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188476357 Chr.6: 34983361-34983361 on Build GRCh38
rs763884374 Chr.6: 34983358-34983358 on Build GRCh38
rs556288300 Chr.6: 34983282-34983282 on Build GRCh38
rs769552537 Chr.6: 34983324-34983324 on Build GRCh38
rs78690508 Chr.6: 34983235-34983235 on Build GRCh38
rs747238008 Chr.6: 34983396-34983396 on Build GRCh38
rs766191563 Chr.6: 34983372-34983372 on Build GRCh38
rs76989831 Chr.6: 34983247-34983247 on Build GRCh38
rs750316121 Chr.6: 34983367-34983367 on Build GRCh38
rs775905379 Chr.6: 34983442-34983442 on Build GRCh38
rs774451073 Chr.6: 34983313-34983313 on Build GRCh38
rs375655597 Chr.6: 34983274-34983274 on Build GRCh38
rs897640126 Chr.6: 34983402-34983402 on Build GRCh38
rs770185043 Chr.6: 34983309-34983309 on Build GRCh38
rs1050178157 Chr.6: 34983364-34983364 on Build GRCh38
rs746378415 Chr.6: 34983287-34983287 on Build GRCh38
rs935557129 Chr.6: 34983270-34983270 on Build GRCh38
rs577896304 Chr.6: 34983424-34983424 on Build GRCh38
rs950900339 Chr.6: 34983280-34983280 on Build GRCh38
rs767526743 Chr.6: 34983436-34983437 on Build GRCh38
rs946364801 Chr.6: 34983337-34983337 on Build GRCh38
rs756062743 Chr.6: 34983371-34983371 on Build GRCh38
rs537664963 Chr.6: 34983277-34983277 on Build GRCh38
rs184886150 Chr.6: 34983256-34983256 on Build GRCh38
rs928249955 Chr.6: 34983269-34983269 on Build GRCh38
rs770247820 Chr.6: 34983430-34983430 on Build GRCh38
rs753615952 Chr.6: 34983374-34983374 on Build GRCh38
rs143407701 Chr.6: 34983285-34983285 on Build GRCh38
rs746385328 Chr.6: 34983429-34983429 on Build GRCh38
rs777932537 Chr.6: 34983384-34983384 on Build GRCh38
rs775787406 Chr.6: 34983326-34983326 on Build GRCh38
rs761903319 Chr.6: 34983365-34983366 on Build GRCh38
rs749616806 Chr.6: 34983444-34983444 on Build GRCh38
rs777130643 Chr.6: 34983284-34983284 on Build GRCh38
rs113792575 Chr.6: 34983434-34983434 on Build GRCh38
rs146179923 Chr.6: 34983344-34983344 on Build GRCh38
rs748388761 Chr.6: 34983255-34983255 on Build GRCh38
rs376166372 Chr.6: 34983440-34983440 on Build GRCh38
rs767137276 Chr.6: 34983365-34983365 on Build GRCh38
rs762667924 Chr.6: 34983332-34983332 on Build GRCh38
rs757271314 Chr.6: 34983407-34983407 on Build GRCh38
rs920945612 Chr.6: 34983398-34983398 on Build GRCh38
rs777545979 Chr.6: 34983376-34983376 on Build GRCh38
rs755868611 Chr.6: 34983244-34983244 on Build GRCh38

View Full Product Details