Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26838208-26838461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS1
Entrez Gene ID
9510
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 1
Gene Aliases
C3-C5, METH1
UniGene
Hs.643357
Gene Chromosome Location
Chr.21: 26836287-26845409 on Build GRCh38
Transcripts
NM_006988.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6094405 c.2204+2T>C p.? Chr.21: 26838437-26838437 on Build GRCh38
444316 c.2215C>A p.H739N Chr.21: 26838268-26838268 on Build GRCh38
4930482 c.2274G>A p.Q758Q Chr.21: 26838209-26838209 on Build GRCh38
6593765 c.2270A>C p.N757T Chr.21: 26838213-26838213 on Build GRCh38
6593757 c.2253C>T p.I751I Chr.21: 26838230-26838230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748093865 Chr.21: 26838299-26838299 on Build GRCh38
rs200052788 Chr.21: 26838258-26838258 on Build GRCh38
rs229042 Chr.21: 26838338-26838338 on Build GRCh38
rs761317711 Chr.21: 26838309-26838310 on Build GRCh38
rs951612731 Chr.21: 26838369-26838369 on Build GRCh38
rs770581245 Chr.21: 26838311-26838311 on Build GRCh38
rs758579925 Chr.21: 26838283-26838283 on Build GRCh38
rs200305587 Chr.21: 26838427-26838427 on Build GRCh38
rs762654704 Chr.21: 26838229-26838229 on Build GRCh38
rs754800494 Chr.21: 26838297-26838297 on Build GRCh38
rs890973394 Chr.21: 26838248-26838248 on Build GRCh38
rs964010286 Chr.21: 26838411-26838411 on Build GRCh38
rs758711789 Chr.21: 26838415-26838415 on Build GRCh38
rs2830550 Chr.21: 26838374-26838374 on Build GRCh38
rs780168461 Chr.21: 26838289-26838289 on Build GRCh38
rs772793999 Chr.21: 26838301-26838301 on Build GRCh38
rs767987172 Chr.21: 26838256-26838258 on Build GRCh38
rs8129724 Chr.21: 26838418-26838418 on Build GRCh38
rs113656634 Chr.21: 26838383-26838383 on Build GRCh38
rs759270807 Chr.21: 26838314-26838314 on Build GRCh38
rs146009181 Chr.21: 26838253-26838253 on Build GRCh38
rs747731086 Chr.21: 26838305-26838306 on Build GRCh38
rs760979769 Chr.21: 26838237-26838237 on Build GRCh38
rs765737157 Chr.21: 26838409-26838409 on Build GRCh38
rs750481824 Chr.21: 26838270-26838270 on Build GRCh38
rs748757489 Chr.21: 26838306-26838306 on Build GRCh38
rs1041916407 Chr.21: 26838240-26838240 on Build GRCh38
rs144090766 Chr.21: 26838230-26838230 on Build GRCh38
rs777210328 Chr.21: 26838433-26838436 on Build GRCh38
rs999240155 Chr.21: 26838233-26838233 on Build GRCh38
rs778661716 Chr.21: 26838282-26838283 on Build GRCh38
rs139582306 Chr.21: 26838259-26838259 on Build GRCh38
rs781174993 Chr.21: 26838298-26838298 on Build GRCh38
rs190846802 Chr.21: 26838312-26838312 on Build GRCh38
rs777033530 Chr.21: 26838396-26838396 on Build GRCh38
rs765284680 Chr.21: 26838268-26838268 on Build GRCh38
rs577135154 Chr.21: 26838425-26838425 on Build GRCh38
rs771577568 Chr.21: 26838327-26838330 on Build GRCh38
rs55911019 Chr.21: 26838300-26838300 on Build GRCh38
rs111919920 Chr.21: 26838279-26838279 on Build GRCh38
rs772524571 Chr.21: 26838308-26838309 on Build GRCh38
rs372677957 Chr.21: 26838316-26838316 on Build GRCh38
rs762246487 Chr.21: 26838406-26838406 on Build GRCh38
rs745779076 Chr.21: 26838282-26838282 on Build GRCh38
rs1007922867 Chr.21: 26838366-26838366 on Build GRCh38
rs1018024475 Chr.21: 26838297-26838297 on Build GRCh38
rs766528566 Chr.21: 26838434-26838434 on Build GRCh38

View Full Product Details