Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50657891-50658138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC3
Entrez Gene ID
8714
Gene Name
ATP binding cassette subfamily C member 3
Gene Aliases
ABC31, EST90757, MLP2, MOAT-D, MRP3, cMOAT2
UniGene
Hs.463421
Gene Chromosome Location
Chr.17: 50634843-50691704 on Build GRCh38
Transcripts
NM_001144070.1 NM_003786.3 XM_005257763.3 XM_011525422.2 XM_011525423.1 XM_011525425.2 XM_017025265.1 XM_017025266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3192784 c.487-8C>A p.? Chr.17: 50658074-50658074 on Build GRCh38
5926946 c.488G>A p.G163D Chr.17: 50658083-50658083 on Build GRCh38
7180995 c.487-1G>A p.? Chr.17: 50658081-50658081 on Build GRCh38
6284410 c.513C>T p.F171F Chr.17: 50658108-50658108 on Build GRCh38
5926947 c.488G>A p.G163D Chr.17: 50658083-50658083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772682310 Chr.17: 50658068-50658068 on Build GRCh38
rs1038168451 Chr.17: 50657921-50657921 on Build GRCh38
rs561199589 Chr.17: 50657942-50657942 on Build GRCh38
rs915000232 Chr.17: 50657917-50657917 on Build GRCh38
rs774731500 Chr.17: 50658052-50658052 on Build GRCh38
rs960678794 Chr.17: 50658016-50658016 on Build GRCh38
rs772284756 Chr.17: 50658035-50658035 on Build GRCh38
rs141668608 Chr.17: 50657937-50657937 on Build GRCh38
rs771299767 Chr.17: 50658049-50658049 on Build GRCh38
rs763550328 Chr.17: 50658105-50658105 on Build GRCh38
rs144810298 Chr.17: 50658109-50658109 on Build GRCh38
rs922614527 Chr.17: 50658113-50658113 on Build GRCh38
rs931637014 Chr.17: 50657919-50657919 on Build GRCh38
rs775846523 Chr.17: 50658077-50658077 on Build GRCh38
rs111998591 Chr.17: 50657953-50657953 on Build GRCh38
rs781253757 Chr.17: 50657963-50657963 on Build GRCh38
rs11568585 Chr.17: 50658031-50658031 on Build GRCh38
rs375412986 Chr.17: 50657947-50657947 on Build GRCh38
rs978886054 Chr.17: 50658054-50658054 on Build GRCh38
rs890140122 Chr.17: 50658015-50658015 on Build GRCh38
rs964303027 Chr.17: 50658060-50658060 on Build GRCh38
rs529892084 Chr.17: 50658027-50658027 on Build GRCh38
rs1002546176 Chr.17: 50657956-50657956 on Build GRCh38
rs543688416 Chr.17: 50658059-50658059 on Build GRCh38
rs545265916 Chr.17: 50658090-50658090 on Build GRCh38
rs760355365 Chr.17: 50658056-50658057 on Build GRCh38
rs750111001 Chr.17: 50658092-50658092 on Build GRCh38
rs200670648 Chr.17: 50658104-50658104 on Build GRCh38
rs562961331 Chr.17: 50658083-50658083 on Build GRCh38

View Full Product Details