Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240509247-240509520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKMY1
Entrez Gene ID
51281
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
ZMYND13
UniGene
Hs.656615
Gene Chromosome Location
Chr.2: 240468632-240561110 on Build GRCh38
Transcripts
NM_001282771.1 NM_001282780.1 NM_001282781.1 NM_001308375.1 NM_016552.3 NM_017844.3 XM_011511300.2 XM_011511301.1 XM_011511304.2 XM_011511305.2 XM_011511309.2 XM_011511313.2 XM_011511317.1 XM_011511318.1 XM_011511322.1 XM_017004271.1 XM_017004272.1 XM_017004273.1 XM_017004274.1 XM_017004275.1 XM_017004276.1 XM_017004277.1 XM_017004278.1 XM_017004279.1 XM_017004280.1 XM_017004281.1 XM_017004282.1 XM_017004283.1 XM_017004284.1 XM_017004285.1 XM_017004286.1 XM_017004287.1 XM_017004288.1 XM_017004289.1 XM_017004290.1 XM_017004291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3579858 c.2070C>T p.N690N Chr.2: 240509405-240509405 on Build GRCh38
4476746 c.2092C>T p.P698S Chr.2: 240509383-240509383 on Build GRCh38
3579859 c.2065C>T p.P689S Chr.2: 240509410-240509410 on Build GRCh38
1018709 c.2111C>A p.A704D Chr.2: 240509364-240509364 on Build GRCh38
6600224 c.2030A>T p.D677V Chr.2: 240509445-240509445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140085976 Chr.2: 240509453-240509453 on Build GRCh38
rs140218422 Chr.2: 240509405-240509405 on Build GRCh38
rs775214519 Chr.2: 240509427-240509429 on Build GRCh38
rs769448136 Chr.2: 240509356-240509356 on Build GRCh38
rs775665615 Chr.2: 240509404-240509404 on Build GRCh38
rs775892946 Chr.2: 240509465-240509465 on Build GRCh38
rs369695358 Chr.2: 240509495-240509495 on Build GRCh38
rs369011359 Chr.2: 240509369-240509369 on Build GRCh38
rs926157619 Chr.2: 240509284-240509284 on Build GRCh38
rs148776770 Chr.2: 240509420-240509420 on Build GRCh38
rs756036071 Chr.2: 240509390-240509390 on Build GRCh38
rs764149270 Chr.2: 240509408-240509408 on Build GRCh38
rs770020155 Chr.2: 240509462-240509462 on Build GRCh38
rs567941473 Chr.2: 240509483-240509483 on Build GRCh38
rs562344050 Chr.2: 240509277-240509277 on Build GRCh38
rs141877484 Chr.2: 240509355-240509355 on Build GRCh38
rs751497859 Chr.2: 240509341-240509341 on Build GRCh38
rs779594668 Chr.2: 240509363-240509363 on Build GRCh38
rs867616801 Chr.2: 240509427-240509427 on Build GRCh38
rs376241917 Chr.2: 240509488-240509488 on Build GRCh38
rs779941650 Chr.2: 240509431-240509431 on Build GRCh38
rs764308915 Chr.2: 240509485-240509485 on Build GRCh38
rs771425474 Chr.2: 240509439-240509439 on Build GRCh38
rs917407268 Chr.2: 240509317-240509317 on Build GRCh38
rs759845841 Chr.2: 240509394-240509394 on Build GRCh38
rs113537399 Chr.2: 240509381-240509381 on Build GRCh38
rs200198187 Chr.2: 240509382-240509382 on Build GRCh38
rs763984945 Chr.2: 240509477-240509478 on Build GRCh38
rs754213489 Chr.2: 240509310-240509310 on Build GRCh38
rs958938570 Chr.2: 240509298-240509298 on Build GRCh38
rs759744807 Chr.2: 240509311-240509311 on Build GRCh38
rs755843558 Chr.2: 240509422-240509422 on Build GRCh38
rs965703281 Chr.2: 240509282-240509282 on Build GRCh38
rs757155921 Chr.2: 240509412-240509412 on Build GRCh38
rs751654113 Chr.2: 240509410-240509410 on Build GRCh38
rs142090859 Chr.2: 240509437-240509437 on Build GRCh38
rs773835301 Chr.2: 240509303-240509303 on Build GRCh38
rs761303207 Chr.2: 240509308-240509308 on Build GRCh38
rs529387037 Chr.2: 240509302-240509302 on Build GRCh38
rs746432795 Chr.2: 240509455-240509455 on Build GRCh38
rs777876971 Chr.2: 240509332-240509332 on Build GRCh38
rs147534670 Chr.2: 240509391-240509391 on Build GRCh38
rs776985961 Chr.2: 240509392-240509392 on Build GRCh38
rs760502825 Chr.2: 240509449-240509449 on Build GRCh38
rs569268612 Chr.2: 240509351-240509351 on Build GRCh38
rs181275702 Chr.2: 240509324-240509324 on Build GRCh38
rs758492419 Chr.2: 240509325-240509325 on Build GRCh38
rs757246647 Chr.2: 240509348-240509348 on Build GRCh38
rs372633587 Chr.2: 240509320-240509320 on Build GRCh38
rs535174102 Chr.2: 240509499-240509499 on Build GRCh38
rs570625017 Chr.2: 240509436-240509436 on Build GRCh38
rs373036321 Chr.2: 240509482-240509482 on Build GRCh38
rs377459989 Chr.2: 240509419-240509419 on Build GRCh38
rs766262453 Chr.2: 240509500-240509500 on Build GRCh38
rs748999358 Chr.2: 240509367-240509367 on Build GRCh38
rs940057306 Chr.2: 240509417-240509417 on Build GRCh38
rs765446831 Chr.2: 240509400-240509400 on Build GRCh38

View Full Product Details