Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57624821-57625056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT1
Entrez Gene ID
2583
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
Gene Aliases
GALGT, GALNACT, GalNAc-T, SPG26
UniGene
Hs.159481
Gene Chromosome Location
Chr.12: 57623409-57633239 on Build GRCh38
Transcripts
XM_005268773.4 XM_011538147.2 XM_011538148.2 XM_017019140.1 XM_017019141.1 XM_017019142.1 NM_133489.2

Gene Symbol
SLC26A10
Entrez Gene ID
65012
Gene Name
solute carrier family 26 member 10
Gene Aliases
-
UniGene
Hs.727726
Gene Chromosome Location
Chr.12: 57619910-57626151 on Build GRCh38
Transcripts
XM_005268773.4 XM_011538147.2 XM_011538148.2 XM_017019140.1 XM_017019141.1 XM_017019142.1 NM_133489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747325 c.1316_1324delGGCACCTGG p.H440_G442delHLG Chr.12: 57624954-57624962 on Build GRCh38
1476792 c.1312G>T p.E438* Chr.12: 57624950-57624950 on Build GRCh38
3463889 c.1245G>C p.G415G Chr.12: 57624883-57624883 on Build GRCh38
4971917 c.1238T>C p.V413A Chr.12: 57624876-57624876 on Build GRCh38
3753421 c.1247T>G p.L416R Chr.12: 57624885-57624885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768099375 Chr.12: 57624894-57624894 on Build GRCh38
rs200643704 Chr.12: 57625020-57625020 on Build GRCh38
rs762803022 Chr.12: 57625031-57625031 on Build GRCh38
rs371428499 Chr.12: 57625024-57625024 on Build GRCh38
rs749700507 Chr.12: 57624941-57624941 on Build GRCh38
rs867905332 Chr.12: 57624903-57624903 on Build GRCh38
rs772629189 Chr.12: 57624964-57624964 on Build GRCh38
rs116098306 Chr.12: 57624967-57624967 on Build GRCh38
rs374579926 Chr.12: 57624927-57624927 on Build GRCh38
rs111924104 Chr.12: 57624885-57624885 on Build GRCh38
rs762691121 Chr.12: 57624922-57624922 on Build GRCh38
rs769459695 Chr.12: 57624969-57624969 on Build GRCh38
rs1038666892 Chr.12: 57624880-57624880 on Build GRCh38
rs772815404 Chr.12: 57624974-57624974 on Build GRCh38
rs748371188 Chr.12: 57624846-57624846 on Build GRCh38
rs998237577 Chr.12: 57624985-57624985 on Build GRCh38
rs767557267 Chr.12: 57624929-57624929 on Build GRCh38
rs767428162 Chr.12: 57625015-57625015 on Build GRCh38
rs764803617 Chr.12: 57624905-57624905 on Build GRCh38
rs771583519 Chr.12: 57624883-57624883 on Build GRCh38
rs200645428 Chr.12: 57624928-57624928 on Build GRCh38
rs566212389 Chr.12: 57624895-57624895 on Build GRCh38
rs919364566 Chr.12: 57625028-57625028 on Build GRCh38
rs765224610 Chr.12: 57624968-57624968 on Build GRCh38
rs766139194 Chr.12: 57624988-57624988 on Build GRCh38
rs750768032 Chr.12: 57624961-57624967 on Build GRCh38
rs1029312125 Chr.12: 57624992-57624992 on Build GRCh38
rs766375429 Chr.12: 57624923-57624923 on Build GRCh38
rs776793049 Chr.12: 57624913-57624913 on Build GRCh38
rs1010614419 Chr.12: 57624897-57624897 on Build GRCh38
rs757736001 Chr.12: 57624951-57624951 on Build GRCh38
rs116545530 Chr.12: 57624879-57624879 on Build GRCh38
rs754162745 Chr.12: 57625032-57625032 on Build GRCh38
rs373627993 Chr.12: 57624962-57624962 on Build GRCh38
rs774185205 Chr.12: 57624999-57624999 on Build GRCh38
rs141552765 Chr.12: 57624926-57624926 on Build GRCh38
rs146389619 Chr.12: 57624938-57624938 on Build GRCh38
rs773004821 Chr.12: 57624919-57624919 on Build GRCh38
rs756397561 Chr.12: 57624849-57624849 on Build GRCh38
rs752754911 Chr.12: 57624930-57624930 on Build GRCh38
rs761522498 Chr.12: 57624899-57624899 on Build GRCh38
rs764237731 Chr.12: 57625030-57625030 on Build GRCh38
rs778284257 Chr.12: 57624877-57624877 on Build GRCh38
rs762565548 Chr.12: 57624977-57624977 on Build GRCh38
rs746354800 Chr.12: 57624888-57624888 on Build GRCh38
rs775089911 Chr.12: 57625008-57625008 on Build GRCh38

View Full Product Details