Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70114143-70114370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM2
Entrez Gene ID
84883
Gene Name
apoptosis inducing factor, mitochondria associated 2
Gene Aliases
AMID, PRG3
UniGene
Hs.650680
Gene Chromosome Location
Chr.10: 70112267-70132934 on Build GRCh38
Transcripts
NM_001198696.1 NM_032797.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2160300 c.1017C>T p.G339G Chr.10: 70114283-70114283 on Build GRCh38
3751918 c.1008C>T p.D336D Chr.10: 70114292-70114292 on Build GRCh38
7296787 c.1088C>T p.T363M Chr.10: 70114212-70114212 on Build GRCh38
7274198 c.1024A>G p.S342G Chr.10: 70114276-70114276 on Build GRCh38
5568097 c.992C>A p.S331Y Chr.10: 70114308-70114308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053495 Chr.10: 70114292-70114292 on Build GRCh38
rs748825232 Chr.10: 70114196-70114196 on Build GRCh38
rs747935542 Chr.10: 70114177-70114177 on Build GRCh38
rs146783519 Chr.10: 70114212-70114212 on Build GRCh38
rs201625501 Chr.10: 70114319-70114319 on Build GRCh38
rs750027306 Chr.10: 70114341-70114341 on Build GRCh38
rs779657402 Chr.10: 70114345-70114345 on Build GRCh38
rs781008253 Chr.10: 70114173-70114173 on Build GRCh38
rs777418200 Chr.10: 70114188-70114188 on Build GRCh38
rs145070950 Chr.10: 70114211-70114211 on Build GRCh38
rs762139673 Chr.10: 70114278-70114278 on Build GRCh38
rs769905584 Chr.10: 70114291-70114291 on Build GRCh38
rs758011663 Chr.10: 70114343-70114343 on Build GRCh38
rs140915557 Chr.10: 70114164-70114164 on Build GRCh38
rs142016168 Chr.10: 70114329-70114329 on Build GRCh38
rs770505998 Chr.10: 70114197-70114197 on Build GRCh38
rs544088598 Chr.10: 70114301-70114301 on Build GRCh38
rs753571116 Chr.10: 70114326-70114326 on Build GRCh38
rs145638743 Chr.10: 70114231-70114231 on Build GRCh38
rs776841617 Chr.10: 70114275-70114275 on Build GRCh38
rs763989866 Chr.10: 70114320-70114320 on Build GRCh38
rs558040718 Chr.10: 70114274-70114274 on Build GRCh38
rs752574364 Chr.10: 70114235-70114235 on Build GRCh38
rs10999144 Chr.10: 70114163-70114163 on Build GRCh38
rs752662377 Chr.10: 70114313-70114313 on Build GRCh38
rs371935916 Chr.10: 70114245-70114245 on Build GRCh38
rs924944848 Chr.10: 70114228-70114228 on Build GRCh38
rs140557384 Chr.10: 70114220-70114220 on Build GRCh38
rs778551581 Chr.10: 70114252-70114252 on Build GRCh38
rs772700059 Chr.10: 70114206-70114206 on Build GRCh38
rs766381889 Chr.10: 70114306-70114306 on Build GRCh38
rs368126515 Chr.10: 70114230-70114230 on Build GRCh38
rs370752237 Chr.10: 70114162-70114162 on Build GRCh38
rs754853382 Chr.10: 70114169-70114169 on Build GRCh38
rs746426577 Chr.10: 70114202-70114202 on Build GRCh38
rs755908795 Chr.10: 70114243-70114243 on Build GRCh38
rs747474561 Chr.10: 70114265-70114265 on Build GRCh38
rs755824143 Chr.10: 70114181-70114181 on Build GRCh38
rs200384144 Chr.10: 70114257-70114257 on Build GRCh38
rs761889088 Chr.10: 70114219-70114219 on Build GRCh38
rs778264558 Chr.10: 70114201-70114201 on Build GRCh38
rs372784129 Chr.10: 70114334-70114334 on Build GRCh38
rs376606751 Chr.10: 70114246-70114246 on Build GRCh38
rs542314841 Chr.10: 70114258-70114258 on Build GRCh38
rs750443319 Chr.10: 70114223-70114223 on Build GRCh38
rs780708284 Chr.10: 70114259-70114259 on Build GRCh38

View Full Product Details