Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92833250-92833517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
-
UniGene
Hs.180946
Gene Chromosome Location
Chr.1: 92832729-92961522 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3

Gene Symbol
RPL5
Entrez Gene ID
6125
Gene Name
ribosomal protein L5
Gene Aliases
DBA6, L5, MSTP030, PPP1R135
UniGene
Hs.532359
Gene Chromosome Location
Chr.1: 92832037-92841924 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4577652 c.21T>C p.V7V Chr.1: 92833406-92833406 on Build GRCh38
6769258 c.73+7T>C p.? Chr.1: 92833465-92833465 on Build GRCh38
391968 c.11_12insTA p.K5fs*15 Chr.1: 92833396-92833397 on Build GRCh38
6570237 c.4-7C>T p.? Chr.1: 92833382-92833382 on Build GRCh38
3401090 c.48C>A p.Y16* Chr.1: 92833433-92833433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764987679 Chr.1: 92833491-92833491 on Build GRCh38
rs373050134 Chr.1: 92833346-92833346 on Build GRCh38
rs777744297 Chr.1: 92833383-92833383 on Build GRCh38
rs142156224 Chr.1: 92833460-92833460 on Build GRCh38
rs746944177 Chr.1: 92833385-92833385 on Build GRCh38
rs114860423 Chr.1: 92833319-92833319 on Build GRCh38
rs750956864 Chr.1: 92833343-92833343 on Build GRCh38
rs571720015 Chr.1: 92833364-92833364 on Build GRCh38
rs943313650 Chr.1: 92833380-92833380 on Build GRCh38
rs752988812 Chr.1: 92833333-92833333 on Build GRCh38
rs777880100 Chr.1: 92833486-92833486 on Build GRCh38
rs772661926 Chr.1: 92833358-92833358 on Build GRCh38
rs774416689 Chr.1: 92833417-92833417 on Build GRCh38
rs14685 Chr.1: 92833448-92833448 on Build GRCh38
rs186600450 Chr.1: 92833360-92833360 on Build GRCh38
rs758256418 Chr.1: 92833495-92833495 on Build GRCh38
rs756108510 Chr.1: 92833483-92833483 on Build GRCh38
rs974273887 Chr.1: 92833377-92833377 on Build GRCh38
rs754413727 Chr.1: 92833490-92833490 on Build GRCh38
rs376462650 Chr.1: 92833359-92833359 on Build GRCh38
rs769877057 Chr.1: 92833391-92833391 on Build GRCh38
rs1032177577 Chr.1: 92833294-92833294 on Build GRCh38
rs555012303 Chr.1: 92833307-92833308 on Build GRCh38
rs369728038 Chr.1: 92833342-92833342 on Build GRCh38
rs761202801 Chr.1: 92833462-92833462 on Build GRCh38
rs762299279 Chr.1: 92833418-92833418 on Build GRCh38
rs121434405 Chr.1: 92833452-92833452 on Build GRCh38
rs778601996 Chr.1: 92833361-92833361 on Build GRCh38
rs779636137 Chr.1: 92833354-92833354 on Build GRCh38
rs1013654956 Chr.1: 92833293-92833293 on Build GRCh38
rs780657111 Chr.1: 92833376-92833376 on Build GRCh38
rs372392388 Chr.1: 92833297-92833297 on Build GRCh38
rs766006888 Chr.1: 92833475-92833475 on Build GRCh38
rs549727048 Chr.1: 92833276-92833276 on Build GRCh38
rs376226148 Chr.1: 92833400-92833400 on Build GRCh38
rs974629918 Chr.1: 92833470-92833470 on Build GRCh38
rs773855879 Chr.1: 92833454-92833454 on Build GRCh38
rs767579888 Chr.1: 92833318-92833318 on Build GRCh38
rs771831213 Chr.1: 92833372-92833372 on Build GRCh38
rs749504844 Chr.1: 92833401-92833401 on Build GRCh38
rs148673599 Chr.1: 92833433-92833433 on Build GRCh38
rs370327480 Chr.1: 92833492-92833492 on Build GRCh38
rs768965548 Chr.1: 92833403-92833403 on Build GRCh38
rs767994083 Chr.1: 92833435-92833435 on Build GRCh38

View Full Product Details