Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161193166-161193437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS4
Entrez Gene ID
9507
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
ADAMTS-2, ADAMTS-4, ADMP-1
UniGene
Hs.211604
Gene Chromosome Location
Chr.1: 161189725-161199080 on Build GRCh38
Transcripts
NM_001320336.1 NM_005099.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5161681 c.1846_1847insC p.Q616fs*48 Chr.1: 161193277-161193278 on Build GRCh38
5095103 c.1793A>T p.K598M Chr.1: 161193331-161193331 on Build GRCh38
5132516 c.1750G>A p.E584K Chr.1: 161193374-161193374 on Build GRCh38
1559885 c.1778G>A p.R593H Chr.1: 161193346-161193346 on Build GRCh38
5003523 c.1783G>T p.D595Y Chr.1: 161193341-161193341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200258484 Chr.1: 161193376-161193376 on Build GRCh38
rs754697059 Chr.1: 161193288-161193288 on Build GRCh38
rs768990385 Chr.1: 161193315-161193315 on Build GRCh38
rs1023316253 Chr.1: 161193285-161193285 on Build GRCh38
rs762045441 Chr.1: 161193397-161193397 on Build GRCh38
rs768415488 Chr.1: 161193212-161193212 on Build GRCh38
rs199819346 Chr.1: 161193318-161193318 on Build GRCh38
rs761593745 Chr.1: 161193243-161193243 on Build GRCh38
rs116198355 Chr.1: 161193231-161193231 on Build GRCh38
rs746213424 Chr.1: 161193374-161193374 on Build GRCh38
rs4233367 Chr.1: 161193247-161193247 on Build GRCh38
rs764200153 Chr.1: 161193236-161193236 on Build GRCh38
rs1033288242 Chr.1: 161193228-161193228 on Build GRCh38
rs776319909 Chr.1: 161193371-161193371 on Build GRCh38
rs775396815 Chr.1: 161193222-161193222 on Build GRCh38
rs371416738 Chr.1: 161193256-161193256 on Build GRCh38
rs780700858 Chr.1: 161193294-161193294 on Build GRCh38
rs772411600 Chr.1: 161193308-161193308 on Build GRCh38
rs374889531 Chr.1: 161193413-161193413 on Build GRCh38
rs758291395 Chr.1: 161193275-161193275 on Build GRCh38
rs931885231 Chr.1: 161193371-161193371 on Build GRCh38
rs779634834 Chr.1: 161193279-161193279 on Build GRCh38
rs751007880 Chr.1: 161193287-161193287 on Build GRCh38
rs371560268 Chr.1: 161193381-161193381 on Build GRCh38
rs760642857 Chr.1: 161193313-161193313 on Build GRCh38
rs762992659 Chr.1: 161193335-161193335 on Build GRCh38
rs766247457 Chr.1: 161193234-161193234 on Build GRCh38
rs529598114 Chr.1: 161193412-161193412 on Build GRCh38
rs139021009 Chr.1: 161193375-161193375 on Build GRCh38
rs751145049 Chr.1: 161193342-161193342 on Build GRCh38
rs577983572 Chr.1: 161193347-161193347 on Build GRCh38
rs779357556 Chr.1: 161193299-161193299 on Build GRCh38
rs143851585 Chr.1: 161193214-161193214 on Build GRCh38
rs746411811 Chr.1: 161193301-161193301 on Build GRCh38
rs775688757 Chr.1: 161193312-161193312 on Build GRCh38
rs368133764 Chr.1: 161193322-161193322 on Build GRCh38
rs760064255 Chr.1: 161193215-161193215 on Build GRCh38
rs202105709 Chr.1: 161193333-161193333 on Build GRCh38
rs776650432 Chr.1: 161193393-161193393 on Build GRCh38
rs767653927 Chr.1: 161193269-161193269 on Build GRCh38
rs754430063 Chr.1: 161193345-161193345 on Build GRCh38
rs148961956 Chr.1: 161193298-161193298 on Build GRCh38
rs1000756366 Chr.1: 161193223-161193223 on Build GRCh38
rs141348315 Chr.1: 161193368-161193368 on Build GRCh38
rs142227379 Chr.1: 161193278-161193278 on Build GRCh38
rs765047810 Chr.1: 161193331-161193331 on Build GRCh38
rs780697413 Chr.1: 161193346-161193346 on Build GRCh38
rs769980910 Chr.1: 161193403-161193403 on Build GRCh38
rs753708382 Chr.1: 161193238-161193238 on Build GRCh38
rs892341257 Chr.1: 161193314-161193314 on Build GRCh38
rs749731245 Chr.1: 161193296-161193296 on Build GRCh38
rs373332448 Chr.1: 161193207-161193207 on Build GRCh38
rs752405036 Chr.1: 161193353-161193353 on Build GRCh38
rs557704452 Chr.1: 161193339-161193339 on Build GRCh38
rs143700956 Chr.1: 161193354-161193354 on Build GRCh38
rs768620886 Chr.1: 161193377-161193377 on Build GRCh38
rs145319567 Chr.1: 161193248-161193248 on Build GRCh38

View Full Product Details