Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113387930-113388167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
UniGene
Hs.448473
Gene Chromosome Location
Chr.11: 113386014-113400418 on Build GRCh38
Transcripts
NM_178510.1 XM_011542736.2 XM_011542737.2 XM_011542738.2 XM_017017475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6433966 c.173C>A p.P58H Chr.11: 113388057-113388057 on Build GRCh38
225321 c.119_120GG>AA p.R40>? Chr.11: 113388003-113388004 on Build GRCh38
1604115 c.74G>T p.W25L Chr.11: 113387958-113387958 on Build GRCh38
5580217 c.139G>A p.E47K Chr.11: 113388023-113388023 on Build GRCh38
6252948 c.145G>T p.A49S Chr.11: 113388029-113388029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756250479 Chr.11: 113388062-113388062 on Build GRCh38
rs963184117 Chr.11: 113387960-113387960 on Build GRCh38
rs142610545 Chr.11: 113387977-113387978 on Build GRCh38
rs879145580 Chr.11: 113387953-113387953 on Build GRCh38
rs1022786023 Chr.11: 113387962-113387962 on Build GRCh38
rs781499658 Chr.11: 113388053-113388053 on Build GRCh38
rs537006404 Chr.11: 113388022-113388022 on Build GRCh38
rs773765126 Chr.11: 113387956-113387956 on Build GRCh38
rs775048985 Chr.11: 113387989-113387989 on Build GRCh38
rs752893588 Chr.11: 113388057-113388057 on Build GRCh38
rs746175705 Chr.11: 113388088-113388088 on Build GRCh38
rs771309001 Chr.11: 113387984-113387984 on Build GRCh38
rs980114929 Chr.11: 113388042-113388042 on Build GRCh38
rs905690204 Chr.11: 113388140-113388140 on Build GRCh38
rs770430581 Chr.11: 113387954-113387954 on Build GRCh38
rs181883517 Chr.11: 113388065-113388065 on Build GRCh38
rs369803292 Chr.11: 113388074-113388074 on Build GRCh38
rs554233180 Chr.11: 113388000-113388000 on Build GRCh38
rs866481043 Chr.11: 113388021-113388021 on Build GRCh38
rs905718104 Chr.11: 113387965-113387965 on Build GRCh38
rs768361670 Chr.11: 113388092-113388092 on Build GRCh38
rs371871655 Chr.11: 113388055-113388055 on Build GRCh38
rs562593885 Chr.11: 113388124-113388124 on Build GRCh38
rs771306482 Chr.11: 113388077-113388077 on Build GRCh38
rs776249480 Chr.11: 113388098-113388098 on Build GRCh38
rs760355089 Chr.11: 113387995-113387995 on Build GRCh38
rs751405790 Chr.11: 113388117-113388117 on Build GRCh38
rs769430211 Chr.11: 113388112-113388112 on Build GRCh38
rs779360079 Chr.11: 113388084-113388084 on Build GRCh38
rs567763343 Chr.11: 113388019-113388019 on Build GRCh38
rs776121430 Chr.11: 113387998-113387998 on Build GRCh38
rs780054479 Chr.11: 113388114-113388114 on Build GRCh38
rs867141063 Chr.11: 113387978-113387978 on Build GRCh38
rs111789052 Chr.11: 113388040-113388040 on Build GRCh38
rs201008406 Chr.11: 113387997-113387997 on Build GRCh38
rs963558639 Chr.11: 113388137-113388137 on Build GRCh38
rs751262148 Chr.11: 113388025-113388025 on Build GRCh38
rs368483324 Chr.11: 113388011-113388011 on Build GRCh38
rs749672480 Chr.11: 113387959-113387959 on Build GRCh38
rs747507046 Chr.11: 113387979-113387994 on Build GRCh38
rs545702309 Chr.11: 113388099-113388099 on Build GRCh38
rs765213302 Chr.11: 113388003-113388003 on Build GRCh38
rs553234761 Chr.11: 113388100-113388100 on Build GRCh38
rs1021319869 Chr.11: 113388033-113388033 on Build GRCh38

View Full Product Details