Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51969940-51970213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD14B
Entrez Gene ID
84836
Gene Name
abhydrolase domain containing 14B
Gene Aliases
CIB, HEL-S-299
UniGene
Hs.420796
Gene Chromosome Location
Chr.3: 51968510-51974630 on Build GRCh38
Transcripts
NM_001146314.1 NM_001254753.1 NM_032750.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269238 c.437C>A p.A146D Chr.3: 51969953-51969953 on Build GRCh38
6165014 c.328C>T p.P110S Chr.3: 51970068-51970068 on Build GRCh38
6202465 c.324C>A p.I108I Chr.3: 51970072-51970072 on Build GRCh38
6269237 c.443C>A p.A148D Chr.3: 51969953-51969953 on Build GRCh38
6590842 c.380C>T p.P127L Chr.3: 51970010-51970010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199563943 Chr.3: 51970155-51970155 on Build GRCh38
rs760136798 Chr.3: 51970041-51970041 on Build GRCh38
rs759213906 Chr.3: 51970053-51970053 on Build GRCh38
rs769965203 Chr.3: 51970010-51970010 on Build GRCh38
rs757439017 Chr.3: 51969996-51969996 on Build GRCh38
rs776168850 Chr.3: 51970046-51970046 on Build GRCh38
rs1010712314 Chr.3: 51970068-51970068 on Build GRCh38
rs778115467 Chr.3: 51970170-51970171 on Build GRCh38
rs780459873 Chr.3: 51970014-51970014 on Build GRCh38
rs775290445 Chr.3: 51970081-51970081 on Build GRCh38
rs150276203 Chr.3: 51970146-51970146 on Build GRCh38
rs751507728 Chr.3: 51970085-51970085 on Build GRCh38
rs770396612 Chr.3: 51970042-51970042 on Build GRCh38
rs745377614 Chr.3: 51970187-51970187 on Build GRCh38
rs755366698 Chr.3: 51970145-51970145 on Build GRCh38
rs373373105 Chr.3: 51970081-51970082 on Build GRCh38
rs1024471782 Chr.3: 51970065-51970065 on Build GRCh38
rs764987211 Chr.3: 51970183-51970183 on Build GRCh38
rs756271218 Chr.3: 51970140-51970140 on Build GRCh38
rs576435421 Chr.3: 51970009-51970009 on Build GRCh38
rs749615200 Chr.3: 51970131-51970131 on Build GRCh38
rs367593230 Chr.3: 51970114-51970114 on Build GRCh38
rs781209309 Chr.3: 51970010-51970024 on Build GRCh38
rs543830840 Chr.3: 51970082-51970082 on Build GRCh38
rs994002623 Chr.3: 51970055-51970055 on Build GRCh38
rs34359997 Chr.3: 51970054-51970055 on Build GRCh38
rs749617726 Chr.3: 51970028-51970028 on Build GRCh38
rs751636953 Chr.3: 51969993-51969993 on Build GRCh38
rs781458494 Chr.3: 51969997-51969997 on Build GRCh38
rs764867980 Chr.3: 51970067-51970067 on Build GRCh38
rs755199192 Chr.3: 51969988-51969990 on Build GRCh38
rs565123236 Chr.3: 51970087-51970087 on Build GRCh38
rs774861827 Chr.3: 51970031-51970031 on Build GRCh38
rs202084346 Chr.3: 51970134-51970134 on Build GRCh38
rs267599887 Chr.3: 51970084-51970084 on Build GRCh38
rs756291732 Chr.3: 51970115-51970115 on Build GRCh38
rs772627435 Chr.3: 51970172-51970172 on Build GRCh38
rs555016805 Chr.3: 51970002-51970002 on Build GRCh38
rs1041819947 Chr.3: 51970079-51970079 on Build GRCh38
rs951091453 Chr.3: 51970156-51970156 on Build GRCh38
rs368664051 Chr.3: 51970030-51970030 on Build GRCh38

View Full Product Details