Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130847225-130847498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY8
Entrez Gene ID
114
Gene Name
adenylate cyclase 8
Gene Aliases
AC8, ADCY3, HBAC1
UniGene
Hs.591859
Gene Chromosome Location
Chr.8: 130780300-131041604 on Build GRCh38
Transcripts
NM_001115.2 XM_005250769.3 XM_006716501.3 XM_017013006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3924630 c.2502+1G>A p.? Chr.8: 130847423-130847423 on Build GRCh38
3645083 c.2465C>T p.S822F Chr.8: 130847461-130847461 on Build GRCh38
3645082 c.2492C>T p.S831F Chr.8: 130847434-130847434 on Build GRCh38
4828178 c.2428G>C p.D810H Chr.8: 130847498-130847498 on Build GRCh38
6595026 c.2465C>A p.S822Y Chr.8: 130847461-130847461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541712490 Chr.8: 130847343-130847343 on Build GRCh38
rs570409152 Chr.8: 130847274-130847274 on Build GRCh38
rs544635095 Chr.8: 130847469-130847469 on Build GRCh38
rs1011731908 Chr.8: 130847417-130847417 on Build GRCh38
rs572567016 Chr.8: 130847356-130847356 on Build GRCh38
rs371352550 Chr.8: 130847391-130847391 on Build GRCh38
rs995742021 Chr.8: 130847263-130847266 on Build GRCh38
rs939722048 Chr.8: 130847336-130847336 on Build GRCh38
rs992601985 Chr.8: 130847434-130847434 on Build GRCh38
rs772079946 Chr.8: 130847474-130847474 on Build GRCh38
rs915765293 Chr.8: 130847281-130847281 on Build GRCh38
rs11997802 Chr.8: 130847378-130847378 on Build GRCh38
rs77429727 Chr.8: 130847455-130847455 on Build GRCh38
rs778119367 Chr.8: 130847428-130847428 on Build GRCh38
rs991328841 Chr.8: 130847255-130847255 on Build GRCh38
rs755145709 Chr.8: 130847397-130847397 on Build GRCh38
rs1018606158 Chr.8: 130847320-130847320 on Build GRCh38
rs368015266 Chr.8: 130847456-130847456 on Build GRCh38
rs761810123 Chr.8: 130847377-130847377 on Build GRCh38
rs753246750 Chr.8: 130847408-130847408 on Build GRCh38
rs886845214 Chr.8: 130847327-130847327 on Build GRCh38
rs34190963 Chr.8: 130847406-130847407 on Build GRCh38
rs747141121 Chr.8: 130847319-130847319 on Build GRCh38
rs371915539 Chr.8: 130847382-130847382 on Build GRCh38
rs1048161568 Chr.8: 130847315-130847315 on Build GRCh38
rs138226842 Chr.8: 130847289-130847291 on Build GRCh38
rs374795546 Chr.8: 130847400-130847400 on Build GRCh38
rs931121637 Chr.8: 130847293-130847293 on Build GRCh38
rs776966258 Chr.8: 130847427-130847427 on Build GRCh38
rs1022164696 Chr.8: 130847451-130847451 on Build GRCh38
rs766557370 Chr.8: 130847389-130847389 on Build GRCh38
rs926528609 Chr.8: 130847404-130847404 on Build GRCh38
rs553089765 Chr.8: 130847392-130847392 on Build GRCh38
rs1040707921 Chr.8: 130847348-130847348 on Build GRCh38
rs1049978321 Chr.8: 130847270-130847270 on Build GRCh38
rs141574722 Chr.8: 130847465-130847465 on Build GRCh38
rs747305191 Chr.8: 130847429-130847429 on Build GRCh38
rs371754667 Chr.8: 130847461-130847461 on Build GRCh38
rs757554640 Chr.8: 130847435-130847435 on Build GRCh38
rs758850690 Chr.8: 130847412-130847412 on Build GRCh38

View Full Product Details