Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10092544-10092817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL6
Entrez Gene ID
83854
Gene Name
angiopoietin like 6
Gene Aliases
AGF, ARP5
UniGene
Hs.712850
Gene Chromosome Location
Chr.19: 10092337-10106194 on Build GRCh38
Transcripts
NM_001321411.1 NM_031917.2 XM_005260091.3 XM_011528347.2 XM_011528348.2 XM_011528349.2 XM_011528350.2 XM_017027347.1 NM_001308277.1 NM_018381.3 XM_011528121.1 XM_017026934.1

Gene Symbol
C19orf66
Entrez Gene ID
55337
Gene Name
chromosome 19 open reading frame 66
Gene Aliases
RyDEN
UniGene
Hs.655613
Gene Chromosome Location
Chr.19: 10086119-10093252 on Build GRCh38
Transcripts
NM_001321411.1 NM_031917.2 XM_005260091.3 XM_011528347.2 XM_011528348.2 XM_011528349.2 XM_011528350.2 XM_017027347.1 NM_001308277.1 NM_018381.3 XM_011528121.1 XM_017026934.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3148548 c.1247delG p.G416fs*53 Chr.19: 10092755-10092755 on Build GRCh38
3148544 c.1322G>A p.R441H Chr.19: 10092680-10092680 on Build GRCh38
6454035 c.1396C>T p.R466W Chr.19: 10092606-10092606 on Build GRCh38
710270 c.1374C>G p.L458L Chr.19: 10092628-10092628 on Build GRCh38
6599667 c.1239C>G p.Y413* Chr.19: 10092763-10092763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042668667 Chr.19: 10092658-10092658 on Build GRCh38
rs540641348 Chr.19: 10092586-10092586 on Build GRCh38
rs138536720 Chr.19: 10092700-10092700 on Build GRCh38
rs371039233 Chr.19: 10092790-10092790 on Build GRCh38
rs764228595 Chr.19: 10092588-10092588 on Build GRCh38
rs745849563 Chr.19: 10092795-10092795 on Build GRCh38
rs971074409 Chr.19: 10092568-10092568 on Build GRCh38
rs141047417 Chr.19: 10092642-10092642 on Build GRCh38
rs45600037 Chr.19: 10092686-10092686 on Build GRCh38
rs780477052 Chr.19: 10092618-10092618 on Build GRCh38
rs967214296 Chr.19: 10092605-10092605 on Build GRCh38
rs774364842 Chr.19: 10092657-10092657 on Build GRCh38
rs376237087 Chr.19: 10092727-10092727 on Build GRCh38
rs150323414 Chr.19: 10092599-10092599 on Build GRCh38
rs769478685 Chr.19: 10092714-10092714 on Build GRCh38
rs770526734 Chr.19: 10092725-10092725 on Build GRCh38
rs149485604 Chr.19: 10092697-10092697 on Build GRCh38
rs765443316 Chr.19: 10092787-10092787 on Build GRCh38
rs754053602 Chr.19: 10092589-10092589 on Build GRCh38
rs757328885 Chr.19: 10092594-10092594 on Build GRCh38
rs759281865 Chr.19: 10092666-10092666 on Build GRCh38
rs751387207 Chr.19: 10092576-10092576 on Build GRCh38
rs915446668 Chr.19: 10092637-10092637 on Build GRCh38
rs377498859 Chr.19: 10092759-10092759 on Build GRCh38
rs771366968 Chr.19: 10092787-10092788 on Build GRCh38
rs771827712 Chr.19: 10092739-10092739 on Build GRCh38
rs548381762 Chr.19: 10092740-10092740 on Build GRCh38
rs767831752 Chr.19: 10092774-10092774 on Build GRCh38
rs748813179 Chr.19: 10092721-10092721 on Build GRCh38
rs777600596 Chr.19: 10092788-10092788 on Build GRCh38
rs765446156 Chr.19: 10092747-10092747 on Build GRCh38
rs372960335 Chr.19: 10092758-10092758 on Build GRCh38
rs910276492 Chr.19: 10092677-10092677 on Build GRCh38
rs375422459 Chr.19: 10092639-10092639 on Build GRCh38
rs756878062 Chr.19: 10092791-10092791 on Build GRCh38
rs769022609 Chr.19: 10092624-10092624 on Build GRCh38
rs756481281 Chr.19: 10092785-10092785 on Build GRCh38
rs745784687 Chr.19: 10092728-10092728 on Build GRCh38
rs778479876 Chr.19: 10092793-10092793 on Build GRCh38
rs138002045 Chr.19: 10092696-10092696 on Build GRCh38
rs766348181 Chr.19: 10092565-10092565 on Build GRCh38
rs201519431 Chr.19: 10092630-10092630 on Build GRCh38
rs146282548 Chr.19: 10092756-10092756 on Build GRCh38
rs781592060 Chr.19: 10092706-10092706 on Build GRCh38
rs772847357 Chr.19: 10092647-10092647 on Build GRCh38
rs760328366 Chr.19: 10092745-10092745 on Build GRCh38
rs141813004 Chr.19: 10092715-10092715 on Build GRCh38
rs369707503 Chr.19: 10092680-10092680 on Build GRCh38
rs747374952 Chr.19: 10092619-10092619 on Build GRCh38
rs753123569 Chr.19: 10092777-10092777 on Build GRCh38
rs754045862 Chr.19: 10092681-10092681 on Build GRCh38
rs750994390 Chr.19: 10092606-10092606 on Build GRCh38
rs200825170 Chr.19: 10092648-10092648 on Build GRCh38
rs201518909 Chr.19: 10092687-10092687 on Build GRCh38
rs747825274 Chr.19: 10092713-10092713 on Build GRCh38
rs45584433 Chr.19: 10092773-10092773 on Build GRCh38
rs771005223 Chr.19: 10092652-10092652 on Build GRCh38
rs150235799 Chr.19: 10092699-10092699 on Build GRCh38
rs759925335 Chr.19: 10092787-10092790 on Build GRCh38
rs777219035 Chr.19: 10092786-10092787 on Build GRCh38
rs879928072 Chr.19: 10092724-10092724 on Build GRCh38

View Full Product Details