Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1012288-1012527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM259
Entrez Gene ID
91304
Gene Name
transmembrane protein 259
Gene Aliases
ASBABP1, C19orf6, MBRL, MEMBRALIN, R32184_3
UniGene
Hs.515003
Gene Chromosome Location
Chr.19: 1009651-1021169 on Build GRCh38
Transcripts
NM_001033026.1 NM_033420.3 XM_005259675.2 XM_005259676.2 XM_005259677.2 XM_005259678.2 XM_017027457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233123 c.662G>A p.R221H Chr.19: 1012519-1012519 on Build GRCh38
5714391 c.718G>A p.D240N Chr.19: 1012463-1012463 on Build GRCh38
4424137 c.676A>C p.T226P Chr.19: 1012505-1012505 on Build GRCh38
5516797 c.700G>A p.V234I Chr.19: 1012481-1012481 on Build GRCh38
1398337 c.669G>A p.S223S Chr.19: 1012512-1012512 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774112698 Chr.19: 1012495-1012495 on Build GRCh38
rs774059547 Chr.19: 1012398-1012398 on Build GRCh38
rs112038545 Chr.19: 1012403-1012403 on Build GRCh38
rs371166014 Chr.19: 1012422-1012422 on Build GRCh38
rs752365131 Chr.19: 1012335-1012335 on Build GRCh38
rs767739162 Chr.19: 1012378-1012379 on Build GRCh38
rs1047761262 Chr.19: 1012348-1012348 on Build GRCh38
rs756751166 Chr.19: 1012370-1012370 on Build GRCh38
rs1027683067 Chr.19: 1012395-1012395 on Build GRCh38
rs1009507476 Chr.19: 1012506-1012506 on Build GRCh38
rs550768513 Chr.19: 1012408-1012408 on Build GRCh38
rs200974669 Chr.19: 1012375-1012375 on Build GRCh38
rs759362772 Chr.19: 1012317-1012317 on Build GRCh38
rs750844247 Chr.19: 1012380-1012382 on Build GRCh38
rs768307881 Chr.19: 1012374-1012375 on Build GRCh38
rs761046351 Chr.19: 1012394-1012396 on Build GRCh38
rs2240162 Chr.19: 1012380-1012380 on Build GRCh38
rs757724979 Chr.19: 1012481-1012481 on Build GRCh38
rs1050975566 Chr.19: 1012482-1012482 on Build GRCh38
rs574528801 Chr.19: 1012324-1012324 on Build GRCh38
rs1038192167 Chr.19: 1012363-1012363 on Build GRCh38
rs775422963 Chr.19: 1012440-1012440 on Build GRCh38
rs747141704 Chr.19: 1012418-1012418 on Build GRCh38
rs1009965255 Chr.19: 1012327-1012327 on Build GRCh38
rs2240163 Chr.19: 1012382-1012382 on Build GRCh38
rs112926823 Chr.19: 1012417-1012417 on Build GRCh38
rs913645254 Chr.19: 1012320-1012320 on Build GRCh38
rs531080154 Chr.19: 1012346-1012346 on Build GRCh38
rs1033777968 Chr.19: 1012386-1012386 on Build GRCh38
rs752997278 Chr.19: 1012415-1012415 on Build GRCh38
rs761515645 Chr.19: 1012502-1012502 on Build GRCh38
rs769652274 Chr.19: 1012385-1012385 on Build GRCh38
rs751046032 Chr.19: 1012362-1012362 on Build GRCh38
rs779104710 Chr.19: 1012375-1012375 on Build GRCh38
rs563541958 Chr.19: 1012336-1012336 on Build GRCh38
rs542927585 Chr.19: 1012379-1012379 on Build GRCh38
rs887850049 Chr.19: 1012349-1012349 on Build GRCh38
rs763833190 Chr.19: 1012396-1012396 on Build GRCh38
rs532250254 Chr.19: 1012405-1012405 on Build GRCh38
rs767742806 Chr.19: 1012467-1012467 on Build GRCh38
rs766807744 Chr.19: 1012417-1012430 on Build GRCh38
rs112874324 Chr.19: 1012413-1012413 on Build GRCh38
rs757298087 Chr.19: 1012421-1012421 on Build GRCh38
rs781093344 Chr.19: 1012423-1012423 on Build GRCh38
rs755335889 Chr.19: 1012374-1012374 on Build GRCh38
rs767074289 Chr.19: 1012505-1012505 on Build GRCh38
rs979167277 Chr.19: 1012383-1012383 on Build GRCh38
rs11672715 Chr.19: 1012441-1012441 on Build GRCh38
rs528124943 Chr.19: 1012381-1012381 on Build GRCh38
rs71339227 Chr.19: 1012380-1012382 on Build GRCh38
rs762698818 Chr.19: 1012392-1012392 on Build GRCh38
rs754397046 Chr.19: 1012406-1012406 on Build GRCh38
rs745829742 Chr.19: 1012425-1012425 on Build GRCh38
rs979564517 Chr.19: 1012330-1012330 on Build GRCh38
rs775087860 Chr.19: 1012387-1012387 on Build GRCh38
rs1054163032 Chr.19: 1012322-1012322 on Build GRCh38
rs369310236 Chr.19: 1012426-1012426 on Build GRCh38
rs748427560 Chr.19: 1012378-1012378 on Build GRCh38
rs374777533 Chr.19: 1012451-1012451 on Build GRCh38
rs780593136 Chr.19: 1012373-1012373 on Build GRCh38

View Full Product Details