Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44568998-44569229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX56
Entrez Gene ID
54606
Gene Name
DEAD-box helicase 56
Gene Aliases
DDX21, DDX26, NOH61
UniGene
Hs.654762
Gene Chromosome Location
Chr.7: 44565417-44574538 on Build GRCh38
Transcripts
NM_001257189.1 NM_019082.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287119 c.1285C>T p.R429C Chr.7: 44569138-44569138 on Build GRCh38
6641985 c.1288T>C p.C430R Chr.7: 44569135-44569135 on Build GRCh38
7364009 c.1256G>A p.R419Q Chr.7: 44569167-44569167 on Build GRCh38
6641990 c.1279C>T p.R427C Chr.7: 44569144-44569144 on Build GRCh38
4164355 c.1263G>A p.E421E Chr.7: 44569160-44569160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368401037 Chr.7: 44569209-44569209 on Build GRCh38
rs751569454 Chr.7: 44569033-44569033 on Build GRCh38
rs749764967 Chr.7: 44569044-44569044 on Build GRCh38
rs776148179 Chr.7: 44569193-44569193 on Build GRCh38
rs1029772292 Chr.7: 44569176-44569176 on Build GRCh38
rs551772363 Chr.7: 44569143-44569143 on Build GRCh38
rs755511857 Chr.7: 44569178-44569178 on Build GRCh38
rs2289051 Chr.7: 44569119-44569119 on Build GRCh38
rs749598622 Chr.7: 44569166-44569166 on Build GRCh38
rs367859317 Chr.7: 44569154-44569154 on Build GRCh38
rs762853797 Chr.7: 44569129-44569129 on Build GRCh38
rs371732045 Chr.7: 44569162-44569162 on Build GRCh38
rs778023776 Chr.7: 44569043-44569043 on Build GRCh38
rs778101565 Chr.7: 44569163-44569163 on Build GRCh38
rs749397340 Chr.7: 44569197-44569198 on Build GRCh38
rs567558816 Chr.7: 44569032-44569032 on Build GRCh38
rs377117026 Chr.7: 44569027-44569027 on Build GRCh38
rs748383962 Chr.7: 44569036-44569036 on Build GRCh38
rs768098186 Chr.7: 44569192-44569192 on Build GRCh38
rs769307680 Chr.7: 44569207-44569207 on Build GRCh38
rs375642630 Chr.7: 44569104-44569104 on Build GRCh38
rs374634183 Chr.7: 44569096-44569096 on Build GRCh38
rs989160241 Chr.7: 44569103-44569103 on Build GRCh38
rs747695681 Chr.7: 44569194-44569194 on Build GRCh38
rs755038623 Chr.7: 44569153-44569153 on Build GRCh38
rs752922054 Chr.7: 44569159-44569159 on Build GRCh38
rs373752875 Chr.7: 44569144-44569144 on Build GRCh38
rs771314930 Chr.7: 44569054-44569054 on Build GRCh38
rs939189465 Chr.7: 44569140-44569140 on Build GRCh38
rs375143988 Chr.7: 44569168-44569168 on Build GRCh38
rs369351947 Chr.7: 44569034-44569034 on Build GRCh38
rs770350598 Chr.7: 44569098-44569099 on Build GRCh38
rs370998600 Chr.7: 44569068-44569068 on Build GRCh38
rs761470602 Chr.7: 44569117-44569117 on Build GRCh38
rs139810379 Chr.7: 44569167-44569167 on Build GRCh38
rs766212961 Chr.7: 44569130-44569130 on Build GRCh38
rs76437461 Chr.7: 44569186-44569186 on Build GRCh38
rs1047781754 Chr.7: 44569169-44569169 on Build GRCh38
rs373489091 Chr.7: 44569035-44569035 on Build GRCh38
rs776269997 Chr.7: 44569123-44569140 on Build GRCh38
rs188278121 Chr.7: 44569086-44569086 on Build GRCh38
rs376757079 Chr.7: 44569039-44569039 on Build GRCh38
rs570150670 Chr.7: 44569151-44569151 on Build GRCh38
rs760165359 Chr.7: 44569093-44569093 on Build GRCh38

View Full Product Details