Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122216504-122216765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIABLO
Entrez Gene ID
56616
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
DFNA64, SMAC
UniGene
Hs.169611
Gene Chromosome Location
Chr.12: 122207662-122227534 on Build GRCh38
Transcripts
NM_001278302.1 NM_001278303.1 NM_001278304.1 NM_001278342.1 NM_019887.5 NM_138930.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7166807 c.424G>A p.E142K Chr.12: 122216761-122216761 on Build GRCh38
5106992 c.502G>A p.A168T Chr.12: 122216509-122216509 on Build GRCh38
1322691 c.459G>A p.L153L Chr.12: 122216552-122216552 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs919112780 Chr.12: 122216612-122216612 on Build GRCh38
rs376774043 Chr.12: 122216551-122216551 on Build GRCh38
rs994617815 Chr.12: 122216537-122216537 on Build GRCh38
rs754040723 Chr.12: 122216617-122216617 on Build GRCh38
rs770624006 Chr.12: 122216718-122216718 on Build GRCh38
rs753115440 Chr.12: 122216629-122216629 on Build GRCh38
rs199768401 Chr.12: 122216539-122216539 on Build GRCh38
rs763205423 Chr.12: 122216726-122216727 on Build GRCh38
rs553958167 Chr.12: 122216698-122216698 on Build GRCh38
rs772893766 Chr.12: 122216592-122216592 on Build GRCh38
rs964638461 Chr.12: 122216678-122216678 on Build GRCh38
rs760778590 Chr.12: 122216614-122216614 on Build GRCh38
rs767503830 Chr.12: 122216587-122216587 on Build GRCh38
rs761643830 Chr.12: 122216579-122216579 on Build GRCh38
rs770727482 Chr.12: 122216712-122216713 on Build GRCh38
rs183090944 Chr.12: 122216567-122216567 on Build GRCh38
rs956490463 Chr.12: 122216734-122216734 on Build GRCh38
rs749902643 Chr.12: 122216722-122216722 on Build GRCh38
rs758764481 Chr.12: 122216721-122216721 on Build GRCh38
rs774045207 Chr.12: 122216736-122216736 on Build GRCh38
rs141116532 Chr.12: 122216622-122216622 on Build GRCh38
rs755662260 Chr.12: 122216739-122216739 on Build GRCh38
rs773101832 Chr.12: 122216595-122216595 on Build GRCh38
rs780506459 Chr.12: 122216529-122216529 on Build GRCh38
rs749069326 Chr.12: 122216620-122216624 on Build GRCh38
rs745376603 Chr.12: 122216535-122216535 on Build GRCh38
rs150291227 Chr.12: 122216674-122216674 on Build GRCh38
rs888418823 Chr.12: 122216640-122216640 on Build GRCh38
rs765502899 Chr.12: 122216623-122216623 on Build GRCh38
rs372537324 Chr.12: 122216601-122216601 on Build GRCh38
rs537707683 Chr.12: 122216616-122216616 on Build GRCh38
rs976987978 Chr.12: 122216683-122216683 on Build GRCh38
rs769722774 Chr.12: 122216560-122216560 on Build GRCh38
rs774270478 Chr.12: 122216569-122216569 on Build GRCh38
rs7952820 Chr.12: 122216656-122216656 on Build GRCh38

View Full Product Details