Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134537076-134537345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP63
Entrez Gene ID
80254
Gene Name
centrosomal protein 63
Gene Aliases
SCKL6
UniGene
Hs.443301
Gene Chromosome Location
Chr.3: 134485733-134629607 on Build GRCh38
Transcripts
NM_001042383.1 NM_001042384.1 NM_001042400.1 NM_025180.3 XM_005247795.4 XM_005247797.3 XM_005247799.3 XM_006713760.3 XM_011513194.2 XM_011513197.2 XM_017007244.1 XM_017007245.1 XM_017007246.1 XM_017007247.1 XM_017007248.1 XM_017007249.1 XM_017007250.1 XM_017007251.1 XM_017007252.1 XM_017007253.1 XM_017007254.1 XM_017007255.1 XM_017007256.1 XM_017007257.1 XM_017007258.1 XM_017007259.1 XM_017007260.1 XM_017007261.1 XM_017007262.1 XM_017007263.1 XM_017007264.1 XM_017007265.1 XM_017007266.1 XM_017007267.1 XM_017007268.1 XM_017007269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3122719 c.442-2A>T p.? Chr.3: 134537153-134537153 on Build GRCh38
4648650 c.500T>C p.V167A Chr.3: 134537213-134537213 on Build GRCh38
243649 c.458C>T p.S153L Chr.3: 134537171-134537171 on Build GRCh38
7278350 c.518A>G p.Q173R Chr.3: 134537231-134537231 on Build GRCh38
243648 c.479G>A p.R160H Chr.3: 134537192-134537192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763184775 Chr.3: 134537313-134537313 on Build GRCh38
rs776766593 Chr.3: 134537272-134537272 on Build GRCh38
rs758803843 Chr.3: 134537211-134537211 on Build GRCh38
rs745713518 Chr.3: 134537188-134537188 on Build GRCh38
rs760883586 Chr.3: 134537115-134537115 on Build GRCh38
rs746885697 Chr.3: 134537253-134537253 on Build GRCh38
rs765578373 Chr.3: 134537139-134537139 on Build GRCh38
rs564376689 Chr.3: 134537301-134537301 on Build GRCh38
rs147553684 Chr.3: 134537191-134537191 on Build GRCh38
rs762027715 Chr.3: 134537204-134537204 on Build GRCh38
rs774195036 Chr.3: 134537187-134537187 on Build GRCh38
rs745623908 Chr.3: 134537250-134537250 on Build GRCh38
rs201316330 Chr.3: 134537162-134537162 on Build GRCh38
rs777174766 Chr.3: 134537171-134537171 on Build GRCh38
rs754047452 Chr.3: 134537130-134537130 on Build GRCh38
rs12634127 Chr.3: 134537317-134537317 on Build GRCh38
rs750790137 Chr.3: 134537141-134537141 on Build GRCh38
rs761850291 Chr.3: 134537283-134537283 on Build GRCh38
rs765441935 Chr.3: 134537207-134537207 on Build GRCh38
rs114108011 Chr.3: 134537268-134537268 on Build GRCh38
rs757441804 Chr.3: 134537136-134537136 on Build GRCh38
rs202055421 Chr.3: 134537192-134537192 on Build GRCh38
rs542172267 Chr.3: 134537198-134537198 on Build GRCh38
rs141971702 Chr.3: 134537217-134537217 on Build GRCh38
rs770951631 Chr.3: 134537282-134537283 on Build GRCh38
rs573165993 Chr.3: 134537169-134537169 on Build GRCh38
rs770497177 Chr.3: 134537180-134537180 on Build GRCh38
rs553384484 Chr.3: 134537165-134537165 on Build GRCh38
rs758895747 Chr.3: 134537161-134537161 on Build GRCh38
rs879200300 Chr.3: 134537208-134537208 on Build GRCh38
rs370527512 Chr.3: 134537299-134537299 on Build GRCh38
rs781656910 Chr.3: 134537221-134537221 on Build GRCh38
rs756924214 Chr.3: 134537235-134537235 on Build GRCh38
rs901425556 Chr.3: 134537284-134537284 on Build GRCh38
rs187494453 Chr.3: 134537227-134537227 on Build GRCh38
rs996432308 Chr.3: 134537239-134537239 on Build GRCh38
rs374262905 Chr.3: 134537104-134537104 on Build GRCh38
rs748916221 Chr.3: 134537172-134537172 on Build GRCh38
rs767968912 Chr.3: 134537318-134537318 on Build GRCh38
rs1036704773 Chr.3: 134537176-134537176 on Build GRCh38
rs372902416 Chr.3: 134537316-134537316 on Build GRCh38
rs148139450 Chr.3: 134537212-134537212 on Build GRCh38
rs778477277 Chr.3: 134537241-134537241 on Build GRCh38
rs771949417 Chr.3: 134537251-134537251 on Build GRCh38

View Full Product Details