Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 20584369-20584620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AY
Entrez Gene ID
9086
Gene Name
eukaryotic translation initiation factor 1A, Y-linked
Gene Aliases
eIF-4C
UniGene
Hs.461178
Gene Chromosome Location
Chr.Y: 20575711-20593154 on Build GRCh38
Transcripts
NM_001278612.1 NM_004681.3

SNP Detail

SNP ID Chromosome Location
rs1048132979 Chr.Y: 20584567-20584567 on Build GRCh38
rs760828225 Chr.Y: 20584535-20584535 on Build GRCh38
rs767785405 Chr.Y: 20584440-20584440 on Build GRCh38
rs754345834 Chr.Y: 20584562-20584562 on Build GRCh38
rs755387941 Chr.Y: 20584568-20584568 on Build GRCh38
rs750548572 Chr.Y: 20584480-20584480 on Build GRCh38
rs767061883 Chr.Y: 20584549-20584549 on Build GRCh38

View Full Product Details