Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50350693-50350926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM186A
Entrez Gene ID
121006
Gene Name
family with sequence similarity 186 member A
Gene Aliases
-
UniGene
Hs.133187
Gene Chromosome Location
Chr.12: 50326854-50396633 on Build GRCh38
Transcripts
NM_001145475.1 XM_006719231.3 XM_011537890.2 XM_011537892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206231 c.6028G>T p.E2010* Chr.12: 50350804-50350804 on Build GRCh38
940330 c.6032C>A p.S2011* Chr.12: 50350800-50350800 on Build GRCh38
4966607 c.6011G>A p.R2004Q Chr.12: 50350821-50350821 on Build GRCh38
1135392 c.6037A>G p.R2013G Chr.12: 50350795-50350795 on Build GRCh38
468487 c.6034T>A p.F2012I Chr.12: 50350798-50350798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536097130 Chr.12: 50350766-50350766 on Build GRCh38
rs1045380040 Chr.12: 50350739-50350739 on Build GRCh38
rs868064144 Chr.12: 50350842-50350842 on Build GRCh38
rs773379306 Chr.12: 50350846-50350846 on Build GRCh38
rs71466904 Chr.12: 50350887-50350887 on Build GRCh38
rs149591363 Chr.12: 50350734-50350734 on Build GRCh38
rs886154287 Chr.12: 50350726-50350726 on Build GRCh38
rs768477883 Chr.12: 50350841-50350841 on Build GRCh38
rs762272218 Chr.12: 50350862-50350862 on Build GRCh38
rs144310021 Chr.12: 50350740-50350740 on Build GRCh38
rs1057338425 Chr.12: 50350821-50350821 on Build GRCh38
rs1010032116 Chr.12: 50350878-50350878 on Build GRCh38
rs893096591 Chr.12: 50350840-50350840 on Build GRCh38
rs905507679 Chr.12: 50350780-50350780 on Build GRCh38
rs1047923777 Chr.12: 50350720-50350720 on Build GRCh38
rs73108300 Chr.12: 50350798-50350798 on Build GRCh38
rs1032918924 Chr.12: 50350807-50350807 on Build GRCh38
rs556022026 Chr.12: 50350855-50350855 on Build GRCh38
rs769778217 Chr.12: 50350785-50350785 on Build GRCh38
rs1000864238 Chr.12: 50350895-50350895 on Build GRCh38
rs566239340 Chr.12: 50350822-50350822 on Build GRCh38
rs183549613 Chr.12: 50350897-50350897 on Build GRCh38
rs905569949 Chr.12: 50350789-50350789 on Build GRCh38
rs1048088471 Chr.12: 50350745-50350745 on Build GRCh38
rs748733450 Chr.12: 50350754-50350755 on Build GRCh38

View Full Product Details