Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81524933-81525206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP12
Entrez Gene ID
646486
Gene Name
fatty acid binding protein 12
Gene Aliases
-
UniGene
Hs.586370
Gene Chromosome Location
Chr.8: 81524673-81590664 on Build GRCh38
Transcripts
NM_001105281.1 XM_005251285.4 XM_006716465.3 XM_011517577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219083 c.422A>C p.*141S Chr.8: 81525047-81525047 on Build GRCh38
6114252 c.398C>A p.S133* Chr.8: 81525071-81525071 on Build GRCh38
3942787 c.419C>G p.S140C Chr.8: 81525050-81525050 on Build GRCh38
4545559 c.380G>A p.R127Q Chr.8: 81525089-81525089 on Build GRCh38
4544103 c.349G>A p.E117K Chr.8: 81525120-81525120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954752867 Chr.8: 81525129-81525129 on Build GRCh38
rs986146785 Chr.8: 81525167-81525167 on Build GRCh38
rs150182306 Chr.8: 81525090-81525090 on Build GRCh38
rs370865801 Chr.8: 81525070-81525070 on Build GRCh38
rs765231107 Chr.8: 81524997-81524997 on Build GRCh38
rs754522064 Chr.8: 81525064-81525064 on Build GRCh38
rs190435213 Chr.8: 81525051-81525051 on Build GRCh38
rs540886808 Chr.8: 81525139-81525139 on Build GRCh38
rs923420855 Chr.8: 81525149-81525149 on Build GRCh38
rs182512224 Chr.8: 81525112-81525112 on Build GRCh38
rs1056930839 Chr.8: 81525061-81525061 on Build GRCh38
rs556911408 Chr.8: 81524998-81524998 on Build GRCh38
rs923878213 Chr.8: 81525135-81525135 on Build GRCh38
rs73275471 Chr.8: 81525162-81525162 on Build GRCh38
rs571622564 Chr.8: 81524961-81524961 on Build GRCh38
rs749605915 Chr.8: 81525075-81525075 on Build GRCh38
rs754539463 Chr.8: 81525157-81525157 on Build GRCh38
rs1042019780 Chr.8: 81524979-81524979 on Build GRCh38
rs118148593 Chr.8: 81525137-81525137 on Build GRCh38
rs1003102279 Chr.8: 81524983-81524983 on Build GRCh38
rs558401182 Chr.8: 81524960-81524960 on Build GRCh38
rs750303552 Chr.8: 81525134-81525134 on Build GRCh38
rs765104699 Chr.8: 81525125-81525125 on Build GRCh38
rs777370625 Chr.8: 81525012-81525012 on Build GRCh38
rs201121910 Chr.8: 81525081-81525081 on Build GRCh38
rs760810562 Chr.8: 81525098-81525098 on Build GRCh38
rs368772586 Chr.8: 81525074-81525074 on Build GRCh38
rs537380648 Chr.8: 81524980-81524980 on Build GRCh38
rs781709156 Chr.8: 81525082-81525082 on Build GRCh38
rs374412876 Chr.8: 81525140-81525140 on Build GRCh38
rs762170047 Chr.8: 81525121-81525121 on Build GRCh38
rs777011733 Chr.8: 81525120-81525120 on Build GRCh38
rs779418749 Chr.8: 81525027-81525027 on Build GRCh38
rs568418174 Chr.8: 81525094-81525094 on Build GRCh38
rs746598180 Chr.8: 81525043-81525043 on Build GRCh38
rs765561681 Chr.8: 81525082-81525083 on Build GRCh38

View Full Product Details