Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36745327-36745588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE5
Entrez Gene ID
57699
Gene Name
copine 5
Gene Aliases
COPN5, CPN5
UniGene
Hs.657869
Gene Chromosome Location
Chr.6: 36740773-36839444 on Build GRCh38
Transcripts
NM_001314018.1 NM_001314019.1 NM_001314020.1 NM_020939.1 XM_005249247.2 XM_011514768.1 XM_011514769.1 XM_011514770.1 XM_011514771.2 XM_011514772.1 XM_011514773.2 XM_011514775.2 XM_017011139.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7272797 c.1328+9C>T p.? Chr.6: 36745379-36745379 on Build GRCh38
451367 c.1266C>T p.S422S Chr.6: 36745450-36745450 on Build GRCh38
1743086 c.1262G>A p.R421H Chr.6: 36745454-36745454 on Build GRCh38
1078573 c.1237G>A p.D413N Chr.6: 36745479-36745479 on Build GRCh38
7207956 c.1284G>T p.L428L Chr.6: 36745432-36745432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543216469 Chr.6: 36745476-36745476 on Build GRCh38
rs554843252 Chr.6: 36745483-36745483 on Build GRCh38
rs866649870 Chr.6: 36745475-36745475 on Build GRCh38
rs377244489 Chr.6: 36745396-36745396 on Build GRCh38
rs748910627 Chr.6: 36745415-36745415 on Build GRCh38
rs372961954 Chr.6: 36745377-36745377 on Build GRCh38
rs147728429 Chr.6: 36745445-36745445 on Build GRCh38
rs895275831 Chr.6: 36745440-36745440 on Build GRCh38
rs539342486 Chr.6: 36745378-36745378 on Build GRCh38
rs771913394 Chr.6: 36745432-36745432 on Build GRCh38
rs773187928 Chr.6: 36745441-36745441 on Build GRCh38
rs781676505 Chr.6: 36745390-36745390 on Build GRCh38
rs760456043 Chr.6: 36745370-36745370 on Build GRCh38
rs554465177 Chr.6: 36745423-36745423 on Build GRCh38
rs754159332 Chr.6: 36745484-36745484 on Build GRCh38
rs761457373 Chr.6: 36745455-36745455 on Build GRCh38
rs764372805 Chr.6: 36745449-36745449 on Build GRCh38
rs1022343458 Chr.6: 36745428-36745428 on Build GRCh38
rs772002184 Chr.6: 36745363-36745363 on Build GRCh38
rs953308406 Chr.6: 36745544-36745544 on Build GRCh38
rs750939871 Chr.6: 36745477-36745477 on Build GRCh38
rs765612901 Chr.6: 36745373-36745373 on Build GRCh38
rs746597357 Chr.6: 36745354-36745354 on Build GRCh38
rs757736623 Chr.6: 36745379-36745379 on Build GRCh38
rs374188493 Chr.6: 36745479-36745479 on Build GRCh38
rs192328387 Chr.6: 36745566-36745566 on Build GRCh38
rs763046 Chr.6: 36745486-36745486 on Build GRCh38
rs146907172 Chr.6: 36745524-36745524 on Build GRCh38
rs369823408 Chr.6: 36745454-36745454 on Build GRCh38
rs377673660 Chr.6: 36745521-36745521 on Build GRCh38
rs778567587 Chr.6: 36745488-36745488 on Build GRCh38
rs747666062 Chr.6: 36745492-36745492 on Build GRCh38
rs756465450 Chr.6: 36745547-36745550 on Build GRCh38
rs774784578 Chr.6: 36745526-36745526 on Build GRCh38
rs779752719 Chr.6: 36745408-36745408 on Build GRCh38
rs565329860 Chr.6: 36745527-36745527 on Build GRCh38
rs747624191 Chr.6: 36745429-36745429 on Build GRCh38
rs1048042687 Chr.6: 36745519-36745519 on Build GRCh38
rs756462231 Chr.6: 36745397-36745397 on Build GRCh38
rs763164595 Chr.6: 36745375-36745375 on Build GRCh38
rs41272180 Chr.6: 36745369-36745369 on Build GRCh38
rs754534056 Chr.6: 36745418-36745418 on Build GRCh38
rs142529265 Chr.6: 36745446-36745446 on Build GRCh38

View Full Product Details