Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167126985-167127216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
UniGene
Hs.632462
Gene Chromosome Location
Chr.1: 167094048-167129165 on Build GRCh38
Transcripts
NM_001080426.1 XM_005245603.3 XM_011510146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6055116 c.2018_2019GG>TT p.G673V Chr.1: 167127149-167127150 on Build GRCh38
3863414 c.2020G>A p.D674N Chr.1: 167127151-167127151 on Build GRCh38
4532550 c.1895G>A p.R632K Chr.1: 167127026-167127026 on Build GRCh38
3376823 c.1944G>A p.A648A Chr.1: 167127075-167127075 on Build GRCh38
4763404 c.1993G>A p.A665T Chr.1: 167127124-167127124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759664430 Chr.1: 167127169-167127171 on Build GRCh38
rs747125220 Chr.1: 167127074-167127074 on Build GRCh38
rs757597404 Chr.1: 167127055-167127055 on Build GRCh38
rs750689944 Chr.1: 167127065-167127065 on Build GRCh38
rs769487453 Chr.1: 167127087-167127087 on Build GRCh38
rs747943857 Chr.1: 167127029-167127029 on Build GRCh38
rs145537197 Chr.1: 167127064-167127064 on Build GRCh38
rs773047471 Chr.1: 167127094-167127094 on Build GRCh38
rs750886063 Chr.1: 167127118-167127118 on Build GRCh38
rs202017285 Chr.1: 167127038-167127038 on Build GRCh38
rs76205550 Chr.1: 167127158-167127158 on Build GRCh38
rs771500645 Chr.1: 167127166-167127166 on Build GRCh38
rs372973596 Chr.1: 167127156-167127156 on Build GRCh38
rs758753192 Chr.1: 167127071-167127071 on Build GRCh38
rs770607161 Chr.1: 167127104-167127104 on Build GRCh38
rs920927738 Chr.1: 167127058-167127058 on Build GRCh38
rs370561353 Chr.1: 167127184-167127184 on Build GRCh38
rs781275928 Chr.1: 167127145-167127145 on Build GRCh38
rs549179166 Chr.1: 167127032-167127032 on Build GRCh38
rs767017134 Chr.1: 167127109-167127109 on Build GRCh38
rs976666073 Chr.1: 167127014-167127014 on Build GRCh38
rs762325833 Chr.1: 167127114-167127114 on Build GRCh38
rs766782601 Chr.1: 167127192-167127192 on Build GRCh38
rs781021697 Chr.1: 167127076-167127076 on Build GRCh38
rs758806552 Chr.1: 167127138-167127138 on Build GRCh38
rs763434444 Chr.1: 167127191-167127191 on Build GRCh38
rs376593703 Chr.1: 167127159-167127159 on Build GRCh38
rs375160987 Chr.1: 167127150-167127150 on Build GRCh38
rs773965871 Chr.1: 167127105-167127105 on Build GRCh38
rs772856160 Chr.1: 167127031-167127031 on Build GRCh38
rs369410112 Chr.1: 167127117-167127117 on Build GRCh38
rs755017675 Chr.1: 167127144-167127144 on Build GRCh38
rs566588987 Chr.1: 167127165-167127165 on Build GRCh38
rs539069832 Chr.1: 167127147-167127147 on Build GRCh38
rs200893444 Chr.1: 167127141-167127141 on Build GRCh38
rs777654054 Chr.1: 167127155-167127155 on Build GRCh38
rs144006881 Chr.1: 167127100-167127100 on Build GRCh38
rs768544982 Chr.1: 167127020-167127020 on Build GRCh38
rs768387689 Chr.1: 167127075-167127075 on Build GRCh38
rs764553686 Chr.1: 167127039-167127039 on Build GRCh38
rs140347809 Chr.1: 167127142-167127142 on Build GRCh38
rs776600431 Chr.1: 167127022-167127022 on Build GRCh38
rs141118809 Chr.1: 167127113-167127113 on Build GRCh38
rs987900395 Chr.1: 167127028-167127028 on Build GRCh38
rs771582809 Chr.1: 167127086-167127086 on Build GRCh38
rs773833411 Chr.1: 167127186-167127186 on Build GRCh38
rs745461356 Chr.1: 167127163-167127163 on Build GRCh38
rs780422021 Chr.1: 167127073-167127073 on Build GRCh38
rs61103736 Chr.1: 167127056-167127056 on Build GRCh38
rs370847886 Chr.1: 167127185-167127185 on Build GRCh38
rs769590965 Chr.1: 167127030-167127030 on Build GRCh38
rs532059690 Chr.1: 167127048-167127048 on Build GRCh38
rs912261228 Chr.1: 167127129-167127129 on Build GRCh38

View Full Product Details