Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31658387-31658656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
UniGene
Hs.368921
Gene Chromosome Location
Chr.1: 31652247-31704167 on Build GRCh38
Transcripts
NM_001856.3 XM_005270481.1 XM_011540722.1 XM_011540723.1 XM_011540724.1 XM_011540726.2 XM_011540727.2 XM_011540728.2 XM_017000338.1 XM_017000339.1 XM_017000340.1 XM_017000341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4931046 c.3984A>C p.P1328P Chr.1: 31658524-31658524 on Build GRCh38
4795443 c.3997C>T p.P1333S Chr.1: 31658511-31658511 on Build GRCh38
5641702 c.3985G>T p.G1329C Chr.1: 31658523-31658523 on Build GRCh38
6239632 c.3992C>A p.P1331H Chr.1: 31658516-31658516 on Build GRCh38
4795442 c.3997C>T p.P1333S Chr.1: 31658511-31658511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772699098 Chr.1: 31658502-31658502 on Build GRCh38
rs371339153 Chr.1: 31658447-31658447 on Build GRCh38
rs757226017 Chr.1: 31658565-31658565 on Build GRCh38
rs60364385 Chr.1: 31658418-31658418 on Build GRCh38
rs767735286 Chr.1: 31658550-31658550 on Build GRCh38
rs868048114 Chr.1: 31658489-31658489 on Build GRCh38
rs775222646 Chr.1: 31658466-31658466 on Build GRCh38
rs374209307 Chr.1: 31658494-31658494 on Build GRCh38
rs768283539 Chr.1: 31658473-31658473 on Build GRCh38
rs751955971 Chr.1: 31658539-31658539 on Build GRCh38
rs561301786 Chr.1: 31658433-31658433 on Build GRCh38
rs1026370028 Chr.1: 31658616-31658616 on Build GRCh38
rs769492223 Chr.1: 31658538-31658538 on Build GRCh38
rs745856920 Chr.1: 31658453-31658453 on Build GRCh38
rs566475071 Chr.1: 31658536-31658536 on Build GRCh38
rs756293418 Chr.1: 31658449-31658449 on Build GRCh38
rs769339168 Chr.1: 31658500-31658500 on Build GRCh38
rs776375012 Chr.1: 31658490-31658490 on Build GRCh38
rs763522147 Chr.1: 31658508-31658508 on Build GRCh38
rs760926747 Chr.1: 31658515-31658515 on Build GRCh38
rs746877223 Chr.1: 31658590-31658590 on Build GRCh38
rs768532199 Chr.1: 31658592-31658592 on Build GRCh38
rs777970305 Chr.1: 31658452-31658452 on Build GRCh38
rs371664162 Chr.1: 31658598-31658598 on Build GRCh38
rs753970176 Chr.1: 31658563-31658563 on Build GRCh38
rs772131141 Chr.1: 31658461-31658461 on Build GRCh38
rs530246679 Chr.1: 31658618-31658618 on Build GRCh38
rs899137331 Chr.1: 31658589-31658589 on Build GRCh38
rs746833027 Chr.1: 31658467-31658467 on Build GRCh38
rs746015691 Chr.1: 31658555-31658556 on Build GRCh38
rs778065260 Chr.1: 31658561-31658561 on Build GRCh38
rs747853046 Chr.1: 31658607-31658607 on Build GRCh38
rs1041836133 Chr.1: 31658429-31658429 on Build GRCh38
rs780825286 Chr.1: 31658596-31658596 on Build GRCh38
rs368113432 Chr.1: 31658551-31658551 on Build GRCh38
rs780062412 Chr.1: 31658586-31658586 on Build GRCh38
rs759969362 Chr.1: 31658541-31658541 on Build GRCh38
rs924370907 Chr.1: 31658432-31658432 on Build GRCh38

View Full Product Details