Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92457077-92457350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPN
Entrez Gene ID
54829
Gene Name
asporin
Gene Aliases
OS3, PLAP-1, PLAP1, SLRR1C
UniGene
Hs.435655
Gene Chromosome Location
Chr.9: 92456205-92482562 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1111118 c.1093C>T p.R365C Chr.9: 92457338-92457338 on Build GRCh38
1463585 c.1099_1100delGT p.V367fs*>14 Chr.9: 92457331-92457332 on Build GRCh38
1183807 c.1109G>A p.R370K Chr.9: 92457322-92457322 on Build GRCh38
6498926 c.1101T>G p.V367V Chr.9: 92457330-92457330 on Build GRCh38
753882 c.1083T>C p.P361P Chr.9: 92457348-92457348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746445338 Chr.9: 92457259-92457259 on Build GRCh38
rs368817815 Chr.9: 92457173-92457173 on Build GRCh38
rs138321845 Chr.9: 92457277-92457277 on Build GRCh38
rs771223864 Chr.9: 92457301-92457301 on Build GRCh38
rs4744132 Chr.9: 92457315-92457315 on Build GRCh38
rs918954979 Chr.9: 92457105-92457105 on Build GRCh38
rs748649282 Chr.9: 92457283-92457283 on Build GRCh38
rs749516419 Chr.9: 92457300-92457300 on Build GRCh38
rs929525388 Chr.9: 92457108-92457108 on Build GRCh38
rs779693811 Chr.9: 92457242-92457242 on Build GRCh38
rs943494361 Chr.9: 92457159-92457161 on Build GRCh38
rs770166448 Chr.9: 92457284-92457284 on Build GRCh38
rs773519082 Chr.9: 92457292-92457292 on Build GRCh38
rs992096621 Chr.9: 92457130-92457130 on Build GRCh38
rs1036406249 Chr.9: 92457226-92457226 on Build GRCh38
rs545140869 Chr.9: 92457274-92457274 on Build GRCh38
rs539889035 Chr.9: 92457210-92457210 on Build GRCh38
rs754675611 Chr.9: 92457276-92457276 on Build GRCh38
rs758043075 Chr.9: 92457239-92457239 on Build GRCh38
rs774720272 Chr.9: 92457309-92457309 on Build GRCh38
rs868522113 Chr.9: 92457264-92457264 on Build GRCh38
rs577499813 Chr.9: 92457216-92457216 on Build GRCh38
rs1039221346 Chr.9: 92457171-92457171 on Build GRCh38
rs917430235 Chr.9: 92457198-92457198 on Build GRCh38
rs893863130 Chr.9: 92457187-92457187 on Build GRCh38
rs559223233 Chr.9: 92457129-92457129 on Build GRCh38

View Full Product Details