Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20142459-20142684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA3
Entrez Gene ID
2700
Gene Name
gap junction protein alpha 3
Gene Aliases
CTRCT14, CX46, CZP3
UniGene
Hs.130313
Gene Chromosome Location
Chr.13: 20138252-20161327 on Build GRCh38
Transcripts
NM_021954.3 XM_011535048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617434 c.819G>A p.A273A Chr.13: 20142470-20142470 on Build GRCh38
5690170 c.734C>A p.A245D Chr.13: 20142555-20142555 on Build GRCh38
6520895 c.721G>T p.D241Y Chr.13: 20142568-20142568 on Build GRCh38
6998363 c.653A>G p.N218S Chr.13: 20142636-20142636 on Build GRCh38
4687007 c.649C>T p.L217F Chr.13: 20142640-20142640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753781759 Chr.13: 20142531-20142531 on Build GRCh38
rs553789027 Chr.13: 20142518-20142518 on Build GRCh38
rs780972084 Chr.13: 20142641-20142641 on Build GRCh38
rs757026999 Chr.13: 20142636-20142636 on Build GRCh38
rs746223535 Chr.13: 20142569-20142569 on Build GRCh38
rs768789596 Chr.13: 20142657-20142657 on Build GRCh38
rs1052432471 Chr.13: 20142476-20142476 on Build GRCh38
rs752524182 Chr.13: 20142616-20142616 on Build GRCh38
rs765833599 Chr.13: 20142499-20142499 on Build GRCh38
rs746511873 Chr.13: 20142634-20142634 on Build GRCh38
rs765170052 Chr.13: 20142587-20142592 on Build GRCh38
rs893768668 Chr.13: 20142479-20142479 on Build GRCh38
rs957763650 Chr.13: 20142511-20142511 on Build GRCh38
rs946255316 Chr.13: 20142522-20142522 on Build GRCh38
rs774650137 Chr.13: 20142573-20142573 on Build GRCh38
rs770016009 Chr.13: 20142647-20142647 on Build GRCh38
rs777200073 Chr.13: 20142568-20142568 on Build GRCh38
rs1045773680 Chr.13: 20142545-20142545 on Build GRCh38
rs755929041 Chr.13: 20142515-20142515 on Build GRCh38
rs777503798 Chr.13: 20142552-20142552 on Build GRCh38
rs3751386 Chr.13: 20142570-20142570 on Build GRCh38
rs760051894 Chr.13: 20142480-20142480 on Build GRCh38
rs753680028 Chr.13: 20142587-20142587 on Build GRCh38
rs771424292 Chr.13: 20142566-20142566 on Build GRCh38
rs764035950 Chr.13: 20142571-20142571 on Build GRCh38
rs774363429 Chr.13: 20142658-20142658 on Build GRCh38
rs764418101 Chr.13: 20142509-20142509 on Build GRCh38
rs752926638 Chr.13: 20142517-20142518 on Build GRCh38
rs761959857 Chr.13: 20142574-20142574 on Build GRCh38
rs751695886 Chr.13: 20142512-20142512 on Build GRCh38
rs752882313 Chr.13: 20142507-20142507 on Build GRCh38
rs576519606 Chr.13: 20142579-20142579 on Build GRCh38
rs572212696 Chr.13: 20142539-20142539 on Build GRCh38
rs746276711 Chr.13: 20142643-20142643 on Build GRCh38
rs757100428 Chr.13: 20142525-20142525 on Build GRCh38
rs762921448 Chr.13: 20142508-20142508 on Build GRCh38
rs746584617 Chr.13: 20142560-20142560 on Build GRCh38
rs757939318 Chr.13: 20142617-20142617 on Build GRCh38
rs888960800 Chr.13: 20142506-20142506 on Build GRCh38
rs1000736399 Chr.13: 20142524-20142524 on Build GRCh38
rs199664100 Chr.13: 20142541-20142541 on Build GRCh38
rs544116497 Chr.13: 20142632-20142632 on Build GRCh38
rs754734137 Chr.13: 20142590-20142590 on Build GRCh38
rs765247252 Chr.13: 20142597-20142597 on Build GRCh38
rs747730140 Chr.13: 20142551-20142551 on Build GRCh38
rs749703145 Chr.13: 20142652-20142652 on Build GRCh38

View Full Product Details