Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47221767-47222026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBC3
Entrez Gene ID
27113
Gene Name
BCL2 binding component 3
Gene Aliases
JFY-1, JFY1, PUMA
UniGene
Hs.467020
Gene Chromosome Location
Chr.19: 47220822-47232998 on Build GRCh38
Transcripts
NM_001127240.2 NM_001127241.2 NM_001127242.2 NM_014417.4 XM_006723141.3 XM_011526722.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384941 c.701C>T p.P234L Chr.19: 47221786-47221786 on Build GRCh38
6610107 c.567G>C p.E189D Chr.19: 47221817-47221817 on Build GRCh38
6979151 c.697_733del37 p.R233fs*>17 Chr.19: 47221754-47221790 on Build GRCh38
6610106 c.670G>C p.D224H Chr.19: 47221817-47221817 on Build GRCh38
7116766 c.607G>C p.E203Q Chr.19: 47221880-47221880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756841779 Chr.19: 47221966-47221966 on Build GRCh38
rs759424372 Chr.19: 47221831-47221831 on Build GRCh38
rs535509958 Chr.19: 47222005-47222005 on Build GRCh38
rs780312516 Chr.19: 47221786-47221786 on Build GRCh38
rs748900635 Chr.19: 47221820-47221820 on Build GRCh38
rs906359318 Chr.19: 47221915-47221915 on Build GRCh38
rs775636828 Chr.19: 47221851-47221851 on Build GRCh38
rs778668010 Chr.19: 47221967-47221967 on Build GRCh38
rs12610787 Chr.19: 47221993-47221993 on Build GRCh38
rs763105913 Chr.19: 47221893-47221893 on Build GRCh38
rs571014601 Chr.19: 47221819-47221819 on Build GRCh38
rs370316669 Chr.19: 47221933-47221933 on Build GRCh38
rs751834949 Chr.19: 47221904-47221904 on Build GRCh38
rs747649341 Chr.19: 47221793-47221793 on Build GRCh38
rs765410456 Chr.19: 47221871-47221871 on Build GRCh38
rs200523681 Chr.19: 47221852-47221852 on Build GRCh38
rs537087759 Chr.19: 47222000-47222000 on Build GRCh38
rs755473445 Chr.19: 47221889-47221889 on Build GRCh38
rs772104737 Chr.19: 47221835-47221835 on Build GRCh38
rs752056442 Chr.19: 47221787-47221787 on Build GRCh38
rs1034388452 Chr.19: 47221792-47221792 on Build GRCh38
rs376693948 Chr.19: 47221971-47221971 on Build GRCh38
rs553748375 Chr.19: 47221934-47221934 on Build GRCh38
rs764146082 Chr.19: 47221928-47221929 on Build GRCh38
rs755399433 Chr.19: 47221929-47221929 on Build GRCh38
rs769349350 Chr.19: 47221795-47221795 on Build GRCh38
rs755414185 Chr.19: 47221789-47221789 on Build GRCh38
rs773442047 Chr.19: 47221890-47221890 on Build GRCh38
rs758721081 Chr.19: 47221785-47221785 on Build GRCh38
rs758746299 Chr.19: 47221927-47221928 on Build GRCh38
rs748926102 Chr.19: 47221944-47221944 on Build GRCh38
rs766673586 Chr.19: 47221899-47221899 on Build GRCh38
rs774040467 Chr.19: 47221824-47221824 on Build GRCh38
rs200472266 Chr.19: 47221868-47221868 on Build GRCh38
rs201625694 Chr.19: 47221975-47221975 on Build GRCh38
rs750140089 Chr.19: 47221978-47221978 on Build GRCh38
rs184609559 Chr.19: 47221790-47221790 on Build GRCh38

View Full Product Details