Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32392165-32392426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTCB
Entrez Gene ID
51493
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Gene Aliases
C22orf28, DJ149A16.6, FAAP, HSPC117
UniGene
Hs.474643
Gene Chromosome Location
Chr.22: 32387575-32412287 on Build GRCh38
Transcripts
NM_014306.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751834 c.1402A>G p.M468V Chr.22: 32392248-32392248 on Build GRCh38
6018224 c.1405G>T p.E469* Chr.22: 32392245-32392245 on Build GRCh38
6770167 c.1379G>A p.R460H Chr.22: 32392271-32392271 on Build GRCh38
5524671 c.1318C>T p.R440* Chr.22: 32392332-32392332 on Build GRCh38
419582 c.1330G>A p.D444N Chr.22: 32392320-32392320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745339287 Chr.22: 32392332-32392332 on Build GRCh38
rs149086655 Chr.22: 32392277-32392277 on Build GRCh38
rs767612001 Chr.22: 32392310-32392310 on Build GRCh38
rs759653853 Chr.22: 32392264-32392264 on Build GRCh38
rs765962569 Chr.22: 32392380-32392384 on Build GRCh38
rs748528006 Chr.22: 32392343-32392343 on Build GRCh38
rs755753729 Chr.22: 32392336-32392336 on Build GRCh38
rs560580867 Chr.22: 32392291-32392291 on Build GRCh38
rs771090582 Chr.22: 32392400-32392400 on Build GRCh38
rs773016415 Chr.22: 32392196-32392197 on Build GRCh38
rs760804713 Chr.22: 32392216-32392216 on Build GRCh38
rs769116977 Chr.22: 32392189-32392189 on Build GRCh38
rs373261643 Chr.22: 32392204-32392204 on Build GRCh38
rs756871532 Chr.22: 32392328-32392328 on Build GRCh38
rs145049274 Chr.22: 32392276-32392276 on Build GRCh38
rs370830559 Chr.22: 32392344-32392344 on Build GRCh38
rs927708981 Chr.22: 32392370-32392370 on Build GRCh38
rs773773602 Chr.22: 32392382-32392382 on Build GRCh38
rs139715698 Chr.22: 32392210-32392210 on Build GRCh38
rs13110 Chr.22: 32392261-32392261 on Build GRCh38
rs751540668 Chr.22: 32392320-32392320 on Build GRCh38
rs747481828 Chr.22: 32392389-32392389 on Build GRCh38
rs966513659 Chr.22: 32392306-32392306 on Build GRCh38
rs201549447 Chr.22: 32392240-32392240 on Build GRCh38
rs748472873 Chr.22: 32392208-32392208 on Build GRCh38
rs773652941 Chr.22: 32392212-32392212 on Build GRCh38
rs779690714 Chr.22: 32392341-32392341 on Build GRCh38
rs776127589 Chr.22: 32392390-32392391 on Build GRCh38
rs5749422 Chr.22: 32392202-32392202 on Build GRCh38
rs145947916 Chr.22: 32392286-32392286 on Build GRCh38
rs760264578 Chr.22: 32392293-32392293 on Build GRCh38

View Full Product Details