Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81190780-81191049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP131
Entrez Gene ID
22994
Gene Name
centrosomal protein 131
Gene Aliases
AZ1, AZI1, ZA1
UniGene
Hs.514578
Gene Chromosome Location
Chr.17: 81189593-81222989 on Build GRCh38
Transcripts
NM_001009811.3 NM_001319228.1 NM_001319229.1 NM_014984.3 XM_011524547.2 XM_011524548.2 XM_011524550.2 XM_011524551.2 XM_011524552.2 XM_011524553.2 XM_011524555.2 XM_017024369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6993038 c.2960C>T p.S987F Chr.17: 81190786-81190786 on Build GRCh38
6148424 c.2931G>T p.E977D Chr.17: 81190919-81190919 on Build GRCh38
4664829 c.2856G>T p.K952N Chr.17: 81190994-81190994 on Build GRCh38
3958927 c.2732G>A p.R911Q Chr.17: 81191010-81191010 on Build GRCh38
6609124 c.2831T>G p.L944R Chr.17: 81191019-81191019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376750143 Chr.17: 81190994-81190994 on Build GRCh38
rs748864105 Chr.17: 81190978-81190978 on Build GRCh38
rs968179365 Chr.17: 81190960-81190960 on Build GRCh38
rs747979401 Chr.17: 81190888-81190888 on Build GRCh38
rs111464253 Chr.17: 81191025-81191025 on Build GRCh38
rs371877115 Chr.17: 81190830-81190830 on Build GRCh38
rs773523503 Chr.17: 81191004-81191004 on Build GRCh38
rs201129332 Chr.17: 81190917-81190917 on Build GRCh38
rs774289584 Chr.17: 81191010-81191010 on Build GRCh38
rs1038619870 Chr.17: 81190896-81190896 on Build GRCh38
rs779719426 Chr.17: 81190817-81190817 on Build GRCh38
rs372462542 Chr.17: 81190951-81190951 on Build GRCh38
rs747453171 Chr.17: 81190933-81190933 on Build GRCh38
rs369443701 Chr.17: 81190942-81190942 on Build GRCh38
rs750388036 Chr.17: 81190802-81190802 on Build GRCh38
rs751341646 Chr.17: 81190910-81190910 on Build GRCh38
rs777148388 Chr.17: 81190926-81190926 on Build GRCh38
rs766337709 Chr.17: 81190965-81190965 on Build GRCh38
rs574947068 Chr.17: 81190959-81190959 on Build GRCh38
rs368111334 Chr.17: 81190828-81190828 on Build GRCh38
rs752508629 Chr.17: 81191023-81191023 on Build GRCh38
rs747588950 Chr.17: 81190984-81190984 on Build GRCh38
rs145199588 Chr.17: 81190961-81190961 on Build GRCh38
rs764434115 Chr.17: 81190905-81190906 on Build GRCh38
rs2279922 Chr.17: 81190907-81190907 on Build GRCh38
rs768259064 Chr.17: 81190827-81190827 on Build GRCh38
rs141181839 Chr.17: 81190969-81190969 on Build GRCh38
rs778275214 Chr.17: 81190982-81190982 on Build GRCh38
rs754073140 Chr.17: 81190933-81190941 on Build GRCh38
rs543818414 Chr.17: 81191008-81191008 on Build GRCh38
rs765497684 Chr.17: 81190952-81190952 on Build GRCh38
rs756844720 Chr.17: 81190979-81190979 on Build GRCh38
rs367948576 Chr.17: 81190897-81190897 on Build GRCh38
rs140907954 Chr.17: 81190929-81190929 on Build GRCh38
rs372194453 Chr.17: 81190913-81190913 on Build GRCh38
rs755773021 Chr.17: 81190975-81190975 on Build GRCh38
rs773113154 Chr.17: 81190908-81190908 on Build GRCh38
rs374960782 Chr.17: 81190884-81190884 on Build GRCh38
rs181279245 Chr.17: 81190986-81190986 on Build GRCh38
rs150154684 Chr.17: 81190930-81190930 on Build GRCh38
rs753586761 Chr.17: 81191026-81191026 on Build GRCh38
rs765481340 Chr.17: 81190901-81190901 on Build GRCh38
rs748770958 Chr.17: 81190927-81190927 on Build GRCh38
rs144132764 Chr.17: 81191006-81191006 on Build GRCh38
rs775726351 Chr.17: 81190849-81190849 on Build GRCh38
rs769203913 Chr.17: 81190941-81190941 on Build GRCh38
rs200499712 Chr.17: 81190970-81190970 on Build GRCh38
rs780940898 Chr.17: 81190914-81190914 on Build GRCh38
rs201675408 Chr.17: 81190974-81190974 on Build GRCh38
rs767380193 Chr.17: 81191020-81191020 on Build GRCh38
rs758353951 Chr.17: 81190806-81190806 on Build GRCh38
rs956013703 Chr.17: 81190855-81190855 on Build GRCh38
rs755756902 Chr.17: 81190925-81190925 on Build GRCh38
rs534252268 Chr.17: 81190847-81190847 on Build GRCh38
rs138890235 Chr.17: 81191011-81191011 on Build GRCh38
rs756858197 Chr.17: 81191027-81191027 on Build GRCh38
rs772693038 Chr.17: 81190832-81190832 on Build GRCh38
rs762116519 Chr.17: 81190900-81190900 on Build GRCh38
rs368486448 Chr.17: 81190824-81190824 on Build GRCh38
rs868388461 Chr.17: 81190946-81190946 on Build GRCh38
rs145065823 Chr.17: 81190976-81190976 on Build GRCh38
rs760025603 Chr.17: 81190840-81190840 on Build GRCh38
rs565029423 Chr.17: 81191003-81191003 on Build GRCh38
rs375749739 Chr.17: 81190987-81190987 on Build GRCh38
rs1033039887 Chr.17: 81190837-81190837 on Build GRCh38
rs746761505 Chr.17: 81190823-81190823 on Build GRCh38
rs140597145 Chr.17: 81191030-81191030 on Build GRCh38
rs750452925 Chr.17: 81190904-81190904 on Build GRCh38
rs542859751 Chr.17: 81190967-81190967 on Build GRCh38
rs761065053 Chr.17: 81190858-81190858 on Build GRCh38
rs747767072 Chr.17: 81190829-81190829 on Build GRCh38
rs143785213 Chr.17: 81191031-81191031 on Build GRCh38

View Full Product Details