Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88529187-88529418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX37
Entrez Gene ID
200523
Gene Name
testis expressed 37
Gene Aliases
C2orf51, TSC21
UniGene
Hs.132104
Gene Chromosome Location
Chr.2: 88524623-88529585 on Build GRCh38
Transcripts
NM_152670.2 XM_005264188.3 XM_011532691.1 XM_011532692.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5570483 c.285G>A p.T95T Chr.2: 88529216-88529216 on Build GRCh38
3583907 c.357C>T p.A119A Chr.2: 88529288-88529288 on Build GRCh38
3583908 c.393C>T p.D131D Chr.2: 88529324-88529324 on Build GRCh38
5551532 c.364G>A p.D122N Chr.2: 88529295-88529295 on Build GRCh38
3583906 c.339C>T p.S113S Chr.2: 88529270-88529270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999788702 Chr.2: 88529280-88529280 on Build GRCh38
rs969815006 Chr.2: 88529312-88529312 on Build GRCh38
rs775750048 Chr.2: 88529298-88529298 on Build GRCh38
rs371396915 Chr.2: 88529316-88529316 on Build GRCh38
rs758353248 Chr.2: 88529369-88529369 on Build GRCh38
rs756020430 Chr.2: 88529302-88529302 on Build GRCh38
rs762328344 Chr.2: 88529225-88529225 on Build GRCh38
rs755701690 Chr.2: 88529230-88529230 on Build GRCh38
rs750788492 Chr.2: 88529229-88529229 on Build GRCh38
rs558903626 Chr.2: 88529324-88529324 on Build GRCh38
rs762238279 Chr.2: 88529304-88529304 on Build GRCh38
rs780035327 Chr.2: 88529262-88529262 on Build GRCh38
rs777841441 Chr.2: 88529380-88529380 on Build GRCh38
rs151127650 Chr.2: 88529273-88529273 on Build GRCh38
rs753505258 Chr.2: 88529244-88529244 on Build GRCh38
rs142818036 Chr.2: 88529215-88529215 on Build GRCh38
rs377723036 Chr.2: 88529283-88529283 on Build GRCh38
rs1013720564 Chr.2: 88529356-88529356 on Build GRCh38
rs146594725 Chr.2: 88529362-88529362 on Build GRCh38
rs764352632 Chr.2: 88529376-88529376 on Build GRCh38
rs751586140 Chr.2: 88529386-88529386 on Build GRCh38
rs753135633 Chr.2: 88529290-88529291 on Build GRCh38
rs766037706 Chr.2: 88529238-88529238 on Build GRCh38
rs903827588 Chr.2: 88529276-88529276 on Build GRCh38
rs774324498 Chr.2: 88529303-88529303 on Build GRCh38
rs72846110 Chr.2: 88529261-88529261 on Build GRCh38
rs770144728 Chr.2: 88529210-88529210 on Build GRCh38
rs933847934 Chr.2: 88529392-88529392 on Build GRCh38
rs866187817 Chr.2: 88529341-88529341 on Build GRCh38
rs777510998 Chr.2: 88529253-88529253 on Build GRCh38
rs570774538 Chr.2: 88529257-88529257 on Build GRCh38
rs748052959 Chr.2: 88529263-88529263 on Build GRCh38
rs183643261 Chr.2: 88529226-88529226 on Build GRCh38
rs764764923 Chr.2: 88529355-88529355 on Build GRCh38
rs770691059 Chr.2: 88529287-88529287 on Build GRCh38
rs139598356 Chr.2: 88529223-88529223 on Build GRCh38
rs148580273 Chr.2: 88529330-88529330 on Build GRCh38
rs117419847 Chr.2: 88529300-88529300 on Build GRCh38
rs374289672 Chr.2: 88529271-88529271 on Build GRCh38
rs61745382 Chr.2: 88529329-88529329 on Build GRCh38
rs200208230 Chr.2: 88529337-88529337 on Build GRCh38
rs746814303 Chr.2: 88529274-88529274 on Build GRCh38
rs760983633 Chr.2: 88529296-88529297 on Build GRCh38
rs1046067568 Chr.2: 88529377-88529377 on Build GRCh38
rs764120157 Chr.2: 88529217-88529217 on Build GRCh38
rs767819395 Chr.2: 88529215-88529216 on Build GRCh38
rs759136123 Chr.2: 88529354-88529354 on Build GRCh38
rs763082564 Chr.2: 88529216-88529216 on Build GRCh38
rs972665391 Chr.2: 88529343-88529343 on Build GRCh38
rs897610577 Chr.2: 88529342-88529342 on Build GRCh38

View Full Product Details