Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38175108-38175351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD9
Entrez Gene ID
114781
Gene Name
BTB domain containing 9
Gene Aliases
dJ322I12.1
UniGene
Hs.603858
Gene Chromosome Location
Chr.6: 38168451-38640426 on Build GRCh38
Transcripts
NM_001099272.1 NM_001172418.1 NM_052893.1 NM_152733.2 XM_011514279.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4530638 c.1510G>A p.G504R Chr.6: 38175110-38175110 on Build GRCh38
3076755 c.1712C>T p.S571L Chr.6: 38175112-38175112 on Build GRCh38
1444182 c.1700delA p.N567fs*>46 Chr.6: 38175124-38175124 on Build GRCh38
7191594 c.1702A>G p.S568G Chr.6: 38175122-38175122 on Build GRCh38
1444183 c.1496delA p.N499fs*>46 Chr.6: 38175124-38175124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982017773 Chr.6: 38175298-38175298 on Build GRCh38
rs541285188 Chr.6: 38175303-38175303 on Build GRCh38
rs764145374 Chr.6: 38175228-38175228 on Build GRCh38
rs770243051 Chr.6: 38175209-38175209 on Build GRCh38
rs750185462 Chr.6: 38175232-38175232 on Build GRCh38
rs913494517 Chr.6: 38175328-38175328 on Build GRCh38
rs781036301 Chr.6: 38175134-38175134 on Build GRCh38
rs762062596 Chr.6: 38175230-38175230 on Build GRCh38
rs946326083 Chr.6: 38175332-38175332 on Build GRCh38
rs759837694 Chr.6: 38175203-38175203 on Build GRCh38
rs913554326 Chr.6: 38175244-38175244 on Build GRCh38
rs899218776 Chr.6: 38175325-38175325 on Build GRCh38
rs185296476 Chr.6: 38175217-38175217 on Build GRCh38
rs937821726 Chr.6: 38175313-38175313 on Build GRCh38
rs964897064 Chr.6: 38175166-38175166 on Build GRCh38
rs200093479 Chr.6: 38175229-38175229 on Build GRCh38
rs369547822 Chr.6: 38175231-38175231 on Build GRCh38
rs748768284 Chr.6: 38175165-38175165 on Build GRCh38
rs949064368 Chr.6: 38175276-38175276 on Build GRCh38
rs116296691 Chr.6: 38175320-38175320 on Build GRCh38
rs769082628 Chr.6: 38175161-38175162 on Build GRCh38
rs777024801 Chr.6: 38175199-38175199 on Build GRCh38
rs546205506 Chr.6: 38175329-38175329 on Build GRCh38
rs368325105 Chr.6: 38175154-38175154 on Build GRCh38
rs372322026 Chr.6: 38175168-38175168 on Build GRCh38
rs545181470 Chr.6: 38175285-38175285 on Build GRCh38
rs987784210 Chr.6: 38175235-38175235 on Build GRCh38
rs926573158 Chr.6: 38175301-38175301 on Build GRCh38
rs780164771 Chr.6: 38175141-38175141 on Build GRCh38
rs375215339 Chr.6: 38175305-38175305 on Build GRCh38
rs771690985 Chr.6: 38175191-38175191 on Build GRCh38
rs927397015 Chr.6: 38175251-38175251 on Build GRCh38
rs747826254 Chr.6: 38175183-38175183 on Build GRCh38
rs778375397 Chr.6: 38175172-38175172 on Build GRCh38
rs189799959 Chr.6: 38175319-38175319 on Build GRCh38
rs775976782 Chr.6: 38175212-38175212 on Build GRCh38
rs375888918 Chr.6: 38175136-38175136 on Build GRCh38

View Full Product Details