Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26508361-26508622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN1A1
Entrez Gene ID
696
Gene Name
butyrophilin subfamily 1 member A1
Gene Aliases
BT, BTN, BTN1
UniGene
Hs.153058
Gene Chromosome Location
Chr.6: 26499290-26510425 on Build GRCh38
Transcripts
NM_001732.2 XM_005249340.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2858535 c.1012A>G p.T338A Chr.6: 26508605-26508605 on Build GRCh38
6513825 c.952C>A p.L318I Chr.6: 26508545-26508545 on Build GRCh38
1442690 c.910G>A p.D304N Chr.6: 26508503-26508503 on Build GRCh38
2858534 c.1006G>C p.E336Q Chr.6: 26508599-26508599 on Build GRCh38
1659489 c.988T>C p.S330P Chr.6: 26508581-26508581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901579443 Chr.6: 26508496-26508496 on Build GRCh38
rs900851330 Chr.6: 26508446-26508446 on Build GRCh38
rs138684847 Chr.6: 26508383-26508383 on Build GRCh38
rs777694746 Chr.6: 26508419-26508419 on Build GRCh38
rs765355838 Chr.6: 26508457-26508458 on Build GRCh38
rs567654520 Chr.6: 26508380-26508380 on Build GRCh38
rs754008731 Chr.6: 26508545-26508545 on Build GRCh38
rs536850553 Chr.6: 26508584-26508584 on Build GRCh38
rs756404007 Chr.6: 26508535-26508535 on Build GRCh38
rs778241200 Chr.6: 26508585-26508585 on Build GRCh38
rs1043673462 Chr.6: 26508402-26508403 on Build GRCh38
rs755009512 Chr.6: 26508569-26508569 on Build GRCh38
rs775105638 Chr.6: 26508455-26508455 on Build GRCh38
rs923888474 Chr.6: 26508430-26508430 on Build GRCh38
rs572169690 Chr.6: 26508414-26508414 on Build GRCh38
rs546665045 Chr.6: 26508394-26508394 on Build GRCh38
rs774265636 Chr.6: 26508485-26508485 on Build GRCh38
rs752997989 Chr.6: 26508480-26508482 on Build GRCh38
rs1980600 Chr.6: 26508501-26508501 on Build GRCh38
rs748609018 Chr.6: 26508570-26508570 on Build GRCh38
rs764387639 Chr.6: 26508540-26508543 on Build GRCh38
rs368401858 Chr.6: 26508530-26508530 on Build GRCh38
rs191341719 Chr.6: 26508408-26508408 on Build GRCh38
rs1040065853 Chr.6: 26508490-26508490 on Build GRCh38
rs757074593 Chr.6: 26508505-26508505 on Build GRCh38
rs753825745 Chr.6: 26508477-26508477 on Build GRCh38
rs142696565 Chr.6: 26508543-26508543 on Build GRCh38
rs375023084 Chr.6: 26508473-26508473 on Build GRCh38
rs371101297 Chr.6: 26508463-26508463 on Build GRCh38
rs557646919 Chr.6: 26508519-26508519 on Build GRCh38

View Full Product Details