Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59124760-59125033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM111B
Entrez Gene ID
374393
Gene Name
family with sequence similarity 111 member B
Gene Aliases
CANP, POIKTMP
UniGene
Hs.186579
Gene Chromosome Location
Chr.11: 59107185-59127415 on Build GRCh38
Transcripts
NM_001142703.1 NM_001142704.1 NM_198947.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7258577 c.848T>A p.I283N Chr.11: 59124945-59124945 on Build GRCh38
6658101 c.918C>G p.H306Q Chr.11: 59125015-59125015 on Build GRCh38
1354960 c.841delA p.I283fs*16 Chr.11: 59124938-59124938 on Build GRCh38
5620776 c.706G>T p.G236C Chr.11: 59124803-59124803 on Build GRCh38
329665 c.845_847delAAA p.K282delK Chr.11: 59124942-59124944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021312103 Chr.11: 59124844-59124844 on Build GRCh38
rs756801588 Chr.11: 59124879-59124879 on Build GRCh38
rs765515289 Chr.11: 59124999-59125000 on Build GRCh38
rs781580149 Chr.11: 59124800-59124800 on Build GRCh38
rs766832243 Chr.11: 59124937-59124938 on Build GRCh38
rs145615939 Chr.11: 59124913-59124913 on Build GRCh38
rs745776814 Chr.11: 59124944-59124944 on Build GRCh38
rs150167727 Chr.11: 59124922-59124922 on Build GRCh38
rs183165256 Chr.11: 59124792-59124792 on Build GRCh38
rs200240410 Chr.11: 59124945-59124945 on Build GRCh38
rs759934000 Chr.11: 59124951-59124954 on Build GRCh38
rs35583029 Chr.11: 59124959-59124959 on Build GRCh38
rs372197121 Chr.11: 59124799-59124799 on Build GRCh38
rs748350402 Chr.11: 59124802-59124802 on Build GRCh38
rs761415295 Chr.11: 59125008-59125008 on Build GRCh38
rs760546563 Chr.11: 59124967-59124967 on Build GRCh38
rs761176801 Chr.11: 59124904-59124907 on Build GRCh38
rs773678626 Chr.11: 59124904-59124905 on Build GRCh38
rs747291996 Chr.11: 59124795-59124795 on Build GRCh38
rs780611406 Chr.11: 59124794-59124794 on Build GRCh38
rs769694630 Chr.11: 59124800-59124801 on Build GRCh38
rs766760952 Chr.11: 59124898-59124898 on Build GRCh38
rs778070886 Chr.11: 59124920-59124920 on Build GRCh38
rs753216985 Chr.11: 59124876-59124876 on Build GRCh38
rs770061233 Chr.11: 59124804-59124804 on Build GRCh38
rs370561169 Chr.11: 59124940-59124940 on Build GRCh38
rs763871323 Chr.11: 59124860-59124860 on Build GRCh38
rs774368257 Chr.11: 59124833-59124833 on Build GRCh38
rs987337627 Chr.11: 59124923-59124923 on Build GRCh38
rs368870336 Chr.11: 59124785-59124785 on Build GRCh38
rs974698808 Chr.11: 59124904-59124904 on Build GRCh38
rs771073776 Chr.11: 59124924-59124924 on Build GRCh38
rs534658111 Chr.11: 59124830-59124830 on Build GRCh38
rs774332841 Chr.11: 59124929-59124929 on Build GRCh38
rs762510876 Chr.11: 59124903-59124904 on Build GRCh38
rs749391696 Chr.11: 59124818-59124818 on Build GRCh38
rs199853146 Chr.11: 59124903-59124903 on Build GRCh38
rs760388824 Chr.11: 59124854-59124854 on Build GRCh38
rs754224751 Chr.11: 59124951-59124952 on Build GRCh38
rs755531775 Chr.11: 59124909-59124909 on Build GRCh38
rs763591821 Chr.11: 59124982-59124982 on Build GRCh38
rs775252721 Chr.11: 59124824-59124827 on Build GRCh38
rs768272759 Chr.11: 59124904-59124904 on Build GRCh38
rs375907586 Chr.11: 59124819-59124819 on Build GRCh38
rs756262360 Chr.11: 59124919-59124919 on Build GRCh38
rs201783823 Chr.11: 59124815-59124815 on Build GRCh38
rs891657075 Chr.11: 59124957-59124957 on Build GRCh38
rs753442611 Chr.11: 59124791-59124791 on Build GRCh38
rs767388540 Chr.11: 59124838-59124838 on Build GRCh38
rs948062788 Chr.11: 59124905-59124905 on Build GRCh38
rs200671340 Chr.11: 59124846-59124846 on Build GRCh38
rs775473066 Chr.11: 59124849-59124849 on Build GRCh38
rs775417936 Chr.11: 59124952-59124952 on Build GRCh38
rs763673160 Chr.11: 59124786-59124786 on Build GRCh38

View Full Product Details