Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147864789-147865062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP10
Entrez Gene ID
79658
Gene Name
Rho GTPase activating protein 10
Gene Aliases
GRAF2, PS-GAP, PSGAP
UniGene
Hs.368631
Gene Chromosome Location
Chr.4: 147732063-148072781 on Build GRCh38
Transcripts
NM_024605.3 XM_005263215.3 XM_017008602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183355 c.538G>A p.E180K Chr.4: 147864897-147864897 on Build GRCh38
3714712 c.568G>C p.E190Q Chr.4: 147864927-147864927 on Build GRCh38
4386498 c.565C>T p.Q189* Chr.4: 147864924-147864924 on Build GRCh38
377950 c.538G>T p.E180* Chr.4: 147864897-147864897 on Build GRCh38
4392044 c.585G>T p.E195D Chr.4: 147864944-147864944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770733944 Chr.4: 147864846-147864846 on Build GRCh38
rs965931369 Chr.4: 147865021-147865021 on Build GRCh38
rs755939515 Chr.4: 147864826-147864826 on Build GRCh38
rs374000912 Chr.4: 147864818-147864818 on Build GRCh38
rs756321585 Chr.4: 147864869-147864870 on Build GRCh38
rs769080340 Chr.4: 147864875-147864875 on Build GRCh38
rs777653155 Chr.4: 147864911-147864911 on Build GRCh38
rs755105567 Chr.4: 147864897-147864897 on Build GRCh38
rs780307514 Chr.4: 147864814-147864814 on Build GRCh38
rs1018614694 Chr.4: 147864907-147864907 on Build GRCh38
rs763083973 Chr.4: 147864887-147864887 on Build GRCh38
rs762417695 Chr.4: 147864883-147864883 on Build GRCh38
rs377558707 Chr.4: 147864842-147864842 on Build GRCh38
rs539067385 Chr.4: 147864892-147864892 on Build GRCh38
rs766376034 Chr.4: 147864890-147864890 on Build GRCh38
rs992642938 Chr.4: 147864930-147864930 on Build GRCh38
rs113916624 Chr.4: 147864904-147864904 on Build GRCh38
rs561455545 Chr.4: 147864858-147864858 on Build GRCh38
rs1008593897 Chr.4: 147864877-147864877 on Build GRCh38
rs530272523 Chr.4: 147864896-147864896 on Build GRCh38
rs781596217 Chr.4: 147864820-147864821 on Build GRCh38
rs550420225 Chr.4: 147864995-147864995 on Build GRCh38
rs778962893 Chr.4: 147864848-147864848 on Build GRCh38
rs768156732 Chr.4: 147865006-147865006 on Build GRCh38
rs757987448 Chr.4: 147864982-147864982 on Build GRCh38
rs886941111 Chr.4: 147864847-147864847 on Build GRCh38
rs753116031 Chr.4: 147864836-147864837 on Build GRCh38
rs749269913 Chr.4: 147864843-147864843 on Build GRCh38
rs776814778 Chr.4: 147864881-147864881 on Build GRCh38
rs775350718 Chr.4: 147864870-147864870 on Build GRCh38
rs779050955 Chr.4: 147864967-147864967 on Build GRCh38
rs776320203 Chr.4: 147865008-147865008 on Build GRCh38
rs753614347 Chr.4: 147864921-147864921 on Build GRCh38
rs374918450 Chr.4: 147864972-147864972 on Build GRCh38
rs758445263 Chr.4: 147864813-147864813 on Build GRCh38
rs547956087 Chr.4: 147864853-147864853 on Build GRCh38
rs746698537 Chr.4: 147865002-147865002 on Build GRCh38
rs765708006 Chr.4: 147864884-147864884 on Build GRCh38
rs757052857 Chr.4: 147864926-147864926 on Build GRCh38
rs370541915 Chr.4: 147864871-147864871 on Build GRCh38
rs750912894 Chr.4: 147864885-147864885 on Build GRCh38

View Full Product Details