Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 140754072-140754321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAF
Entrez Gene ID
673
Gene Name
B-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
B-RAF1, B-raf, BRAF1, NS7, RAFB1
UniGene
Hs.550061
Gene Chromosome Location
Chr.7: 140719331-140924764 on Build GRCh38
Transcripts
NM_004333.4 XM_005250045.2 XM_017012558.1 XM_017012559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5406678 c.1695-2A>G p.? Chr.7: 140754235-140754235 on Build GRCh38
364756 c.1722C>A p.H574Q Chr.7: 140754206-140754206 on Build GRCh38
364757 c.1720C>A p.H574N Chr.7: 140754208-140754208 on Build GRCh38
2861428 c.1741A>T p.N581Y Chr.7: 140754187-140754187 on Build GRCh38
6912607 c.1720C>T p.H574Y Chr.7: 140754208-140754208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750735369 Chr.7: 140754172-140754172 on Build GRCh38
rs768059168 Chr.7: 140754163-140754163 on Build GRCh38
rs749538915 Chr.7: 140754197-140754197 on Build GRCh38
rs371218602 Chr.7: 140754279-140754279 on Build GRCh38
rs544780847 Chr.7: 140754173-140754173 on Build GRCh38
rs773887544 Chr.7: 140754285-140754285 on Build GRCh38
rs112847359 Chr.7: 140754129-140754129 on Build GRCh38
rs397507480 Chr.7: 140754233-140754233 on Build GRCh38
rs756501278 Chr.7: 140754177-140754177 on Build GRCh38
rs769640108 Chr.7: 140754256-140754256 on Build GRCh38
rs371142723 Chr.7: 140754269-140754269 on Build GRCh38
rs762308238 Chr.7: 140754144-140754144 on Build GRCh38
rs909262796 Chr.7: 140754108-140754108 on Build GRCh38
rs34568439 Chr.7: 140754136-140754137 on Build GRCh38
rs1057517915 Chr.7: 140754202-140754202 on Build GRCh38
rs374085430 Chr.7: 140754286-140754286 on Build GRCh38
rs869246798 Chr.7: 140754252-140754252 on Build GRCh38
rs200228275 Chr.7: 140754203-140754203 on Build GRCh38
rs775202459 Chr.7: 140754258-140754258 on Build GRCh38
rs762658943 Chr.7: 140754261-140754261 on Build GRCh38
rs763643518 Chr.7: 140754149-140754149 on Build GRCh38
rs563382605 Chr.7: 140754249-140754249 on Build GRCh38
rs757727430 Chr.7: 140754162-140754162 on Build GRCh38
rs755214031 Chr.7: 140754214-140754214 on Build GRCh38
rs397516894 Chr.7: 140754208-140754208 on Build GRCh38
rs180177040 Chr.7: 140754187-140754187 on Build GRCh38
rs199714538 Chr.7: 140754224-140754224 on Build GRCh38
rs189077091 Chr.7: 140754287-140754287 on Build GRCh38
rs889385940 Chr.7: 140754283-140754283 on Build GRCh38
rs375383775 Chr.7: 140754116-140754116 on Build GRCh38
rs397507481 Chr.7: 140754206-140754206 on Build GRCh38
rs869148494 Chr.7: 140754137-140754138 on Build GRCh38
rs751094451 Chr.7: 140754157-140754157 on Build GRCh38
rs780073231 Chr.7: 140754178-140754178 on Build GRCh38
rs748279980 Chr.7: 140754218-140754218 on Build GRCh38

View Full Product Details