Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 116487292-116487565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN2
Entrez Gene ID
23245
Gene Name
astrotactin 2
Gene Aliases
bA67K19.1
UniGene
Hs.601562
Gene Chromosome Location
Chr.9: 116425225-117415039 on Build GRCh38
Transcripts
NM_001184734.1 NM_014010.4 NM_198186.3 NM_198187.3 NM_198188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6347019 c.3342C>A p.Y1114* Chr.9: 116487361-116487361 on Build GRCh38
6606197 c.725G>A p.R242Q Chr.9: 116487434-116487434 on Build GRCh38
3653550 c.3286C>T p.P1096S Chr.9: 116487417-116487417 on Build GRCh38
68964 c.3215_3224del10 p.S1072fs*12 Chr.9: 116487479-116487488 on Build GRCh38
5875425 c.577C>T p.R193* Chr.9: 116487435-116487435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577478546 Chr.9: 116487370-116487370 on Build GRCh38
rs760388560 Chr.9: 116487481-116487481 on Build GRCh38
rs749231349 Chr.9: 116487331-116487331 on Build GRCh38
rs773214659 Chr.9: 116487333-116487333 on Build GRCh38
rs767618025 Chr.9: 116487535-116487535 on Build GRCh38
rs201974916 Chr.9: 116487504-116487504 on Build GRCh38
rs766522381 Chr.9: 116487434-116487434 on Build GRCh38
rs561860594 Chr.9: 116487340-116487340 on Build GRCh38
rs962313321 Chr.9: 116487400-116487400 on Build GRCh38
rs755027418 Chr.9: 116487418-116487418 on Build GRCh38
rs749000872 Chr.9: 116487451-116487451 on Build GRCh38
rs771706204 Chr.9: 116487478-116487478 on Build GRCh38
rs535121691 Chr.9: 116487534-116487534 on Build GRCh38
rs146615956 Chr.9: 116487456-116487456 on Build GRCh38
rs996634449 Chr.9: 116487512-116487512 on Build GRCh38
rs768126115 Chr.9: 116487498-116487498 on Build GRCh38
rs775179334 Chr.9: 116487362-116487362 on Build GRCh38
rs924987556 Chr.9: 116487484-116487484 on Build GRCh38
rs781306907 Chr.9: 116487425-116487425 on Build GRCh38
rs765621808 Chr.9: 116487369-116487369 on Build GRCh38
rs199818695 Chr.9: 116487461-116487461 on Build GRCh38
rs16933591 Chr.9: 116487411-116487411 on Build GRCh38
rs763253094 Chr.9: 116487522-116487522 on Build GRCh38
rs373090315 Chr.9: 116487325-116487325 on Build GRCh38
rs747940015 Chr.9: 116487320-116487320 on Build GRCh38
rs752658890 Chr.9: 116487536-116487536 on Build GRCh38
rs756393988 Chr.9: 116487543-116487543 on Build GRCh38
rs142855762 Chr.9: 116487455-116487455 on Build GRCh38
rs377397516 Chr.9: 116487526-116487526 on Build GRCh38
rs777634484 Chr.9: 116487445-116487445 on Build GRCh38
rs762152883 Chr.9: 116487363-116487363 on Build GRCh38
rs140483683 Chr.9: 116487408-116487408 on Build GRCh38
rs763454702 Chr.9: 116487373-116487373 on Build GRCh38
rs759898304 Chr.9: 116487507-116487507 on Build GRCh38
rs755917557 Chr.9: 116487435-116487435 on Build GRCh38
rs531920780 Chr.9: 116487506-116487506 on Build GRCh38
rs766919952 Chr.9: 116487529-116487529 on Build GRCh38
rs771902290 Chr.9: 116487345-116487345 on Build GRCh38

View Full Product Details