Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24225597-24225850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT5
Entrez Gene ID
2687
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGL, GGT 5, GGT-REL, GGTLA1
UniGene
Hs.437156
Gene Chromosome Location
Chr.22: 24219654-24245142 on Build GRCh38
Transcripts
NM_001099781.2 NM_001099782.2 NM_001302464.1 NM_001302465.1 NM_004121.3 XM_005261557.2 XM_005261558.2 XM_011530133.1 XM_011530134.1 XM_011530135.1 XM_011530136.1 XM_011530137.2 XM_017028768.1 XM_017028769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208149 c.1256G>A p.R419Q Chr.22: 24225626-24225626 on Build GRCh38
3552628 c.1261G>A p.G421S Chr.22: 24225621-24225621 on Build GRCh38
726140 c.1238C>T p.A413V Chr.22: 24225644-24225644 on Build GRCh38
1683565 c.1275_1276insA p.N426fs*87 Chr.22: 24225606-24225607 on Build GRCh38
6297316 c.1238C>A p.A413E Chr.22: 24225644-24225644 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200073930 Chr.22: 24225618-24225618 on Build GRCh38
rs1006560724 Chr.22: 24225711-24225711 on Build GRCh38
rs755235008 Chr.22: 24225619-24225619 on Build GRCh38
rs1048426049 Chr.22: 24225705-24225705 on Build GRCh38
rs189178297 Chr.22: 24225723-24225723 on Build GRCh38
rs374036306 Chr.22: 24225699-24225699 on Build GRCh38
rs1016185568 Chr.22: 24225715-24225715 on Build GRCh38
rs546779082 Chr.22: 24225773-24225773 on Build GRCh38
rs201765477 Chr.22: 24225631-24225631 on Build GRCh38
rs1029261650 Chr.22: 24225695-24225695 on Build GRCh38
rs972045743 Chr.22: 24225722-24225722 on Build GRCh38
rs1014620904 Chr.22: 24225753-24225753 on Build GRCh38
rs529592678 Chr.22: 24225639-24225639 on Build GRCh38
rs999878962 Chr.22: 24225763-24225763 on Build GRCh38
rs916070531 Chr.22: 24225747-24225747 on Build GRCh38
rs368114799 Chr.22: 24225661-24225661 on Build GRCh38
rs748517292 Chr.22: 24225624-24225624 on Build GRCh38
rs1036855208 Chr.22: 24225740-24225740 on Build GRCh38
rs769426895 Chr.22: 24225667-24225667 on Build GRCh38
rs778369692 Chr.22: 24225626-24225626 on Build GRCh38
rs535139256 Chr.22: 24225791-24225791 on Build GRCh38
rs935029942 Chr.22: 24225776-24225776 on Build GRCh38
rs916055461 Chr.22: 24225620-24225620 on Build GRCh38
rs534493457 Chr.22: 24225757-24225757 on Build GRCh38
rs924850065 Chr.22: 24225764-24225764 on Build GRCh38
rs142733225 Chr.22: 24225625-24225625 on Build GRCh38
rs747475402 Chr.22: 24225627-24225627 on Build GRCh38
rs118007626 Chr.22: 24225756-24225756 on Build GRCh38
rs1052063709 Chr.22: 24225777-24225777 on Build GRCh38
rs562982518 Chr.22: 24225691-24225691 on Build GRCh38
rs372565443 Chr.22: 24225676-24225676 on Build GRCh38
rs71318962 Chr.22: 24225735-24225735 on Build GRCh38
rs143000629 Chr.22: 24225814-24225814 on Build GRCh38
rs568226124 Chr.22: 24225781-24225781 on Build GRCh38
rs779215682 Chr.22: 24225622-24225622 on Build GRCh38
rs761779669 Chr.22: 24225668-24225668 on Build GRCh38
rs752816688 Chr.22: 24225701-24225701 on Build GRCh38
rs557389866 Chr.22: 24225822-24225822 on Build GRCh38
rs764492598 Chr.22: 24225741-24225741 on Build GRCh38
rs753960025 Chr.22: 24225690-24225690 on Build GRCh38
rs780218835 Chr.22: 24225629-24225629 on Build GRCh38
rs759245413 Chr.22: 24225641-24225641 on Build GRCh38
rs901486137 Chr.22: 24225785-24225785 on Build GRCh38
rs775196188 Chr.22: 24225660-24225660 on Build GRCh38
rs769456009 Chr.22: 24225644-24225644 on Build GRCh38
rs564017458 Chr.22: 24225664-24225664 on Build GRCh38
rs962061374 Chr.22: 24225718-24225718 on Build GRCh38
rs988291433 Chr.22: 24225824-24225824 on Build GRCh38

View Full Product Details