Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68713858-68714091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2B
Entrez Gene ID
10391
Gene Name
coronin 2B
Gene Aliases
CLIPINC
UniGene
Hs.551213
Gene Chromosome Location
Chr.15: 68518373-68727806 on Build GRCh38
Transcripts
NM_001190456.1 NM_001190457.1 NM_001324014.1 NM_001324015.1 NM_006091.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5454379 c.717G>A p.T239T Chr.15: 68713993-68713993 on Build GRCh38
555890 c.702G>T p.M234I Chr.15: 68713978-68713978 on Build GRCh38
333998 c.669C>A p.H223Q Chr.15: 68713945-68713945 on Build GRCh38
4806859 c.701T>C p.M234T Chr.15: 68713977-68713977 on Build GRCh38
280087 c.726C>A p.V242V Chr.15: 68714002-68714002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751549062 Chr.15: 68713999-68713999 on Build GRCh38
rs752421094 Chr.15: 68714023-68714023 on Build GRCh38
rs553808342 Chr.15: 68713983-68713983 on Build GRCh38
rs773347405 Chr.15: 68714070-68714070 on Build GRCh38
rs939691680 Chr.15: 68713972-68713972 on Build GRCh38
rs754897969 Chr.15: 68714000-68714000 on Build GRCh38
rs932408551 Chr.15: 68713941-68713941 on Build GRCh38
rs777152740 Chr.15: 68714061-68714061 on Build GRCh38
rs750542270 Chr.15: 68713987-68713987 on Build GRCh38
rs886790421 Chr.15: 68713946-68713946 on Build GRCh38
rs575833365 Chr.15: 68714051-68714051 on Build GRCh38
rs939244449 Chr.15: 68714025-68714025 on Build GRCh38
rs865875374 Chr.15: 68714060-68714060 on Build GRCh38
rs144987289 Chr.15: 68713956-68713956 on Build GRCh38
rs762134833 Chr.15: 68713960-68713960 on Build GRCh38
rs1036695628 Chr.15: 68713982-68713982 on Build GRCh38
rs748416317 Chr.15: 68714062-68714062 on Build GRCh38
rs372496796 Chr.15: 68713897-68713897 on Build GRCh38
rs17852400 Chr.15: 68713988-68713988 on Build GRCh38
rs555909560 Chr.15: 68714038-68714038 on Build GRCh38
rs539164836 Chr.15: 68713932-68713932 on Build GRCh38
rs922083476 Chr.15: 68713898-68713898 on Build GRCh38
rs781030685 Chr.15: 68714019-68714019 on Build GRCh38
rs769222104 Chr.15: 68714058-68714058 on Build GRCh38
rs931234018 Chr.15: 68713913-68713913 on Build GRCh38
rs755804205 Chr.15: 68714028-68714028 on Build GRCh38
rs770086605 Chr.15: 68714066-68714066 on Build GRCh38
rs766452912 Chr.15: 68713992-68713992 on Build GRCh38
rs756753941 Chr.15: 68714050-68714050 on Build GRCh38
rs1046692734 Chr.15: 68713926-68713926 on Build GRCh38
rs369781686 Chr.15: 68714048-68714048 on Build GRCh38
rs996347967 Chr.15: 68714057-68714057 on Build GRCh38
rs1048141586 Chr.15: 68713943-68713943 on Build GRCh38
rs762902893 Chr.15: 68713990-68713990 on Build GRCh38

View Full Product Details