Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19521757-19522028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDE1
Entrez Gene ID
51573
Gene Name
glycerophosphodiester phosphodiesterase 1
Gene Aliases
363E6.2, MIR16
UniGene
Hs.512607
Gene Chromosome Location
Chr.16: 19501693-19522145 on Build GRCh38
Transcripts
NM_001324066.1 NM_001324067.1 NM_016641.3 NR_136689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619435 c.184C>T p.R62W Chr.16: 19521781-19521781 on Build GRCh38
6001969 c.20A>T p.Q7L Chr.16: 19521945-19521945 on Build GRCh38
5971681 c.201C>T p.A67A Chr.16: 19521764-19521764 on Build GRCh38
4464370 c.132C>T p.V44V Chr.16: 19521833-19521833 on Build GRCh38
237444 c.72G>T p.V24V Chr.16: 19521893-19521893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745980551 Chr.16: 19521867-19521867 on Build GRCh38
rs778268045 Chr.16: 19521856-19521856 on Build GRCh38
rs759095009 Chr.16: 19521980-19521980 on Build GRCh38
rs754285597 Chr.16: 19521888-19521888 on Build GRCh38
rs572378276 Chr.16: 19521989-19521989 on Build GRCh38
rs771192961 Chr.16: 19521797-19521797 on Build GRCh38
rs768667010 Chr.16: 19521813-19521813 on Build GRCh38
rs746404618 Chr.16: 19521824-19521824 on Build GRCh38
rs374811586 Chr.16: 19521843-19521843 on Build GRCh38
rs554469966 Chr.16: 19521781-19521781 on Build GRCh38
rs757513332 Chr.16: 19521892-19521892 on Build GRCh38
rs778961906 Chr.16: 19521861-19521861 on Build GRCh38
rs958375578 Chr.16: 19521907-19521907 on Build GRCh38
rs757557909 Chr.16: 19521859-19521859 on Build GRCh38
rs776381521 Chr.16: 19521821-19521821 on Build GRCh38
rs756612176 Chr.16: 19521851-19521851 on Build GRCh38
rs761274376 Chr.16: 19521886-19521886 on Build GRCh38
rs867033644 Chr.16: 19521944-19521944 on Build GRCh38
rs143742836 Chr.16: 19521775-19521775 on Build GRCh38
rs749603189 Chr.16: 19521857-19521857 on Build GRCh38
rs558336573 Chr.16: 19521850-19521850 on Build GRCh38
rs750577838 Chr.16: 19521914-19521914 on Build GRCh38
rs758571997 Chr.16: 19521920-19521920 on Build GRCh38
rs764988589 Chr.16: 19521828-19521828 on Build GRCh38
rs772021698 Chr.16: 19521868-19521868 on Build GRCh38
rs756449389 Chr.16: 19521853-19521853 on Build GRCh38
rs768093000 Chr.16: 19521847-19521847 on Build GRCh38
rs554135504 Chr.16: 19521979-19521979 on Build GRCh38
rs762584757 Chr.16: 19521830-19521830 on Build GRCh38
rs1021123155 Chr.16: 19522003-19522003 on Build GRCh38
rs780257381 Chr.16: 19521854-19521854 on Build GRCh38
rs199729378 Chr.16: 19521870-19521870 on Build GRCh38
rs747033594 Chr.16: 19521950-19521950 on Build GRCh38
rs760929197 Chr.16: 19521869-19521869 on Build GRCh38
rs749703548 Chr.16: 19521947-19521947 on Build GRCh38
rs534319317 Chr.16: 19521873-19521873 on Build GRCh38
rs766929737 Chr.16: 19521881-19521881 on Build GRCh38
rs866399210 Chr.16: 19521959-19521959 on Build GRCh38
rs773829743 Chr.16: 19521877-19521877 on Build GRCh38
rs751085725 Chr.16: 19521842-19521842 on Build GRCh38
rs781486541 Chr.16: 19521780-19521780 on Build GRCh38
rs11551690 Chr.16: 19521887-19521887 on Build GRCh38
rs199987552 Chr.16: 19521955-19521955 on Build GRCh38
rs769732063 Chr.16: 19521952-19521952 on Build GRCh38
rs374774754 Chr.16: 19521965-19521965 on Build GRCh38
rs779985406 Chr.16: 19521930-19521930 on Build GRCh38
rs371787862 Chr.16: 19521885-19521885 on Build GRCh38
rs748074496 Chr.16: 19521871-19521871 on Build GRCh38
rs142292419 Chr.16: 19521809-19521809 on Build GRCh38
rs368007979 Chr.16: 19521776-19521776 on Build GRCh38
rs749515587 Chr.16: 19521794-19521794 on Build GRCh38
rs770650017 Chr.16: 19521901-19521901 on Build GRCh38
rs11551689 Chr.16: 19521774-19521774 on Build GRCh38
rs1021438582 Chr.16: 19521964-19521964 on Build GRCh38
rs759137540 Chr.16: 19521878-19521878 on Build GRCh38
rs910844576 Chr.16: 19521988-19521988 on Build GRCh38
rs749107454 Chr.16: 19521956-19521956 on Build GRCh38
rs201638661 Chr.16: 19521836-19521836 on Build GRCh38
rs774202356 Chr.16: 19521945-19521947 on Build GRCh38
rs779243290 Chr.16: 19521897-19521897 on Build GRCh38
rs372769514 Chr.16: 19522001-19522001 on Build GRCh38
rs988218243 Chr.16: 19521960-19521960 on Build GRCh38
rs778179810 Chr.16: 19521789-19521789 on Build GRCh38
rs201477567 Chr.16: 19521801-19521801 on Build GRCh38
rs377212006 Chr.16: 19521876-19521876 on Build GRCh38
rs371574971 Chr.16: 19521787-19521787 on Build GRCh38
rs35567178 Chr.16: 19521872-19521872 on Build GRCh38

View Full Product Details