Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25724459-25724732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J
Entrez Gene ID
522
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6
Gene Aliases
ATP5, ATP5A, ATPM, CF6, F6
UniGene
Hs.246310
Gene Chromosome Location
Chr.21: 25724480-25735654 on Build GRCh38
Transcripts
NM_001003696.1 NM_001003697.1 NM_001003701.1 NM_001003703.1 NM_001320266.1 NM_001320267.1 NM_001685.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4634589 c.315A>G p.K105K Chr.21: 25724652-25724652 on Build GRCh38
5366237 c.315T>C p.D105D Chr.21: 25724676-25724676 on Build GRCh38
7227317 c.290-5C>G p.? Chr.21: 25724682-25724682 on Build GRCh38
4780778 c.335A>G p.E112G Chr.21: 25724656-25724656 on Build GRCh38
6607965 c.290-10delC p.? Chr.21: 25724687-25724687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372728601 Chr.21: 25724586-25724586 on Build GRCh38
rs917291483 Chr.21: 25724600-25724600 on Build GRCh38
rs939429836 Chr.21: 25724562-25724562 on Build GRCh38
rs937457495 Chr.21: 25724556-25724556 on Build GRCh38
rs755692518 Chr.21: 25724692-25724692 on Build GRCh38
rs768771738 Chr.21: 25724638-25724638 on Build GRCh38
rs747217889 Chr.21: 25724635-25724635 on Build GRCh38
rs770432263 Chr.21: 25724703-25724703 on Build GRCh38
rs759141938 Chr.21: 25724592-25724592 on Build GRCh38
rs769613924 Chr.21: 25724681-25724681 on Build GRCh38
rs554635248 Chr.21: 25724542-25724542 on Build GRCh38
rs754271063 Chr.21: 25724597-25724597 on Build GRCh38
rs200700811 Chr.21: 25724604-25724604 on Build GRCh38
rs11539867 Chr.21: 25724649-25724649 on Build GRCh38
rs138359468 Chr.21: 25724643-25724643 on Build GRCh38
rs928021688 Chr.21: 25724507-25724507 on Build GRCh38
rs1036291223 Chr.21: 25724617-25724617 on Build GRCh38
rs757754345 Chr.21: 25724603-25724603 on Build GRCh38
rs73335993 Chr.21: 25724700-25724700 on Build GRCh38
rs369627415 Chr.21: 25724605-25724605 on Build GRCh38
rs540364911 Chr.21: 25724619-25724619 on Build GRCh38
rs373517431 Chr.21: 25724620-25724620 on Build GRCh38
rs780022177 Chr.21: 25724627-25724627 on Build GRCh38
rs370688247 Chr.21: 25724691-25724692 on Build GRCh38
rs926130853 Chr.21: 25724549-25724549 on Build GRCh38
rs1803181 Chr.21: 25724650-25724650 on Build GRCh38
rs377177728 Chr.21: 25724658-25724658 on Build GRCh38
rs994744738 Chr.21: 25724624-25724624 on Build GRCh38
rs201392159 Chr.21: 25724689-25724689 on Build GRCh38
rs372312396 Chr.21: 25724596-25724596 on Build GRCh38

View Full Product Details