Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4274094-4274323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCND2
Entrez Gene ID
894
Gene Name
cyclin D2
Gene Aliases
KIAK0002, MPPH3
UniGene
Hs.376071
Gene Chromosome Location
Chr.12: 4273736-4305356 on Build GRCh38
Transcripts
NM_001759.3 NR_125790.1

Gene Symbol
CCND2-AS1
Entrez Gene ID
103752584
Gene Name
CCND2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.577711
Gene Chromosome Location
Chr.12: 4248767-4276184 on Build GRCh38
Transcripts
NM_001759.3 NR_125790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333234 c.65G>A p.R22Q Chr.12: 4274105-4274105 on Build GRCh38
7313648 c.195G>A p.E65E Chr.12: 4274235-4274235 on Build GRCh38
2066983 c.195+2T>C p.? Chr.12: 4274237-4274237 on Build GRCh38
5003689 c.162C>A p.Y54* Chr.12: 4274202-4274202 on Build GRCh38
1242108 c.137G>A p.C46Y Chr.12: 4274177-4274177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754626676 Chr.12: 4274293-4274293 on Build GRCh38
rs758596420 Chr.12: 4274283-4274283 on Build GRCh38
rs146122734 Chr.12: 4274154-4274154 on Build GRCh38
rs371866493 Chr.12: 4274290-4274290 on Build GRCh38
rs772607278 Chr.12: 4274248-4274248 on Build GRCh38
rs778986120 Chr.12: 4274128-4274128 on Build GRCh38
rs61731957 Chr.12: 4274209-4274209 on Build GRCh38
rs779857983 Chr.12: 4274136-4274136 on Build GRCh38
rs772187536 Chr.12: 4274134-4274134 on Build GRCh38
rs367795095 Chr.12: 4274257-4274257 on Build GRCh38
rs780210978 Chr.12: 4274242-4274243 on Build GRCh38
rs768686427 Chr.12: 4274151-4274151 on Build GRCh38
rs769290616 Chr.12: 4274213-4274213 on Build GRCh38
rs746820501 Chr.12: 4274149-4274149 on Build GRCh38
rs1014244917 Chr.12: 4274244-4274244 on Build GRCh38
rs778984815 Chr.12: 4274277-4274277 on Build GRCh38
rs558500960 Chr.12: 4274256-4274256 on Build GRCh38
rs956736880 Chr.12: 4274137-4274137 on Build GRCh38
rs761064579 Chr.12: 4274264-4274264 on Build GRCh38
rs757774242 Chr.12: 4274269-4274269 on Build GRCh38
rs764578479 Chr.12: 4274265-4274265 on Build GRCh38
rs746798022 Chr.12: 4274284-4274284 on Build GRCh38
rs761395165 Chr.12: 4274199-4274199 on Build GRCh38
rs560920176 Chr.12: 4274262-4274262 on Build GRCh38
rs746879522 Chr.12: 4274243-4274243 on Build GRCh38
rs746043858 Chr.12: 4274131-4274131 on Build GRCh38
rs200011361 Chr.12: 4274241-4274241 on Build GRCh38
rs1009356572 Chr.12: 4274138-4274138 on Build GRCh38
rs372251438 Chr.12: 4274267-4274267 on Build GRCh38
rs541133384 Chr.12: 4274261-4274261 on Build GRCh38
rs750498163 Chr.12: 4274278-4274278 on Build GRCh38

View Full Product Details