Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102933603-102933856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
UniGene
Hs.654634
Gene Chromosome Location
Chr.14: 102932379-103057405 on Build GRCh38
Transcripts
NM_006035.3 XM_005268227.1 XM_005268228.1 XM_005268229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4654722 c.5108G>A p.G1703D Chr.14: 102933740-102933740 on Build GRCh38
3493932 c.5071C>T p.P1691S Chr.14: 102933777-102933777 on Build GRCh38
4846255 c.5078C>G p.S1693C Chr.14: 102933770-102933770 on Build GRCh38
1707899 c.5065_5066CC>TT p.P1689L Chr.14: 102933782-102933783 on Build GRCh38
3711428 c.5021C>T p.T1674I Chr.14: 102933827-102933827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778842513 Chr.14: 102933653-102933653 on Build GRCh38
rs758672847 Chr.14: 102933824-102933824 on Build GRCh38
rs768331940 Chr.14: 102933816-102933816 on Build GRCh38
rs764290721 Chr.14: 102933825-102933825 on Build GRCh38
rs367568433 Chr.14: 102933678-102933678 on Build GRCh38
rs977864022 Chr.14: 102933727-102933727 on Build GRCh38
rs761881673 Chr.14: 102933700-102933700 on Build GRCh38
rs780343468 Chr.14: 102933764-102933765 on Build GRCh38
rs767539948 Chr.14: 102933707-102933707 on Build GRCh38
rs764049477 Chr.14: 102933790-102933790 on Build GRCh38
rs752816236 Chr.14: 102933729-102933729 on Build GRCh38
rs1051753126 Chr.14: 102933634-102933634 on Build GRCh38
rs970445027 Chr.14: 102933704-102933704 on Build GRCh38
rs765191399 Chr.14: 102933809-102933809 on Build GRCh38
rs932077513 Chr.14: 102933632-102933632 on Build GRCh38
rs371638840 Chr.14: 102933763-102933763 on Build GRCh38
rs745757978 Chr.14: 102933665-102933665 on Build GRCh38
rs535677589 Chr.14: 102933820-102933820 on Build GRCh38
rs755007496 Chr.14: 102933721-102933721 on Build GRCh38
rs751989434 Chr.14: 102933832-102933832 on Build GRCh38
rs758823144 Chr.14: 102933744-102933744 on Build GRCh38
rs369647222 Chr.14: 102933808-102933808 on Build GRCh38
rs35239675 Chr.14: 102933745-102933745 on Build GRCh38
rs376636312 Chr.14: 102933682-102933682 on Build GRCh38
rs774458674 Chr.14: 102933699-102933699 on Build GRCh38
rs774336264 Chr.14: 102933769-102933769 on Build GRCh38
rs35064888 Chr.14: 102933666-102933666 on Build GRCh38
rs201614344 Chr.14: 102933686-102933686 on Build GRCh38
rs376036277 Chr.14: 102933781-102933781 on Build GRCh38
rs781663654 Chr.14: 102933748-102933748 on Build GRCh38
rs140640502 Chr.14: 102933747-102933747 on Build GRCh38
rs10139783 Chr.14: 102933648-102933648 on Build GRCh38
rs924097013 Chr.14: 102933739-102933739 on Build GRCh38
rs775226335 Chr.14: 102933764-102933764 on Build GRCh38
rs933824918 Chr.14: 102933805-102933805 on Build GRCh38
rs548033579 Chr.14: 102933691-102933691 on Build GRCh38
rs761685326 Chr.14: 102933775-102933775 on Build GRCh38
rs548479650 Chr.14: 102933661-102933661 on Build GRCh38
rs765510912 Chr.14: 102933728-102933728 on Build GRCh38
rs372216187 Chr.14: 102933664-102933664 on Build GRCh38
rs758646022 Chr.14: 102933716-102933717 on Build GRCh38
rs757553589 Chr.14: 102933835-102933835 on Build GRCh38
rs772966692 Chr.14: 102933799-102933799 on Build GRCh38
rs760303410 Chr.14: 102933765-102933765 on Build GRCh38
rs562965814 Chr.14: 102933645-102933645 on Build GRCh38
rs562551309 Chr.14: 102933761-102933761 on Build GRCh38
rs1051697726 Chr.14: 102933788-102933788 on Build GRCh38
rs762818594 Chr.14: 102933692-102933692 on Build GRCh38
rs767627879 Chr.14: 102933789-102933789 on Build GRCh38

View Full Product Details