Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41690605-41690876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf46
Entrez Gene ID
79640
Gene Name
chromosome 22 open reading frame 46
Gene Aliases
-
UniGene
Hs.517653
Gene Chromosome Location
Chr.22: 41688939-41698136 on Build GRCh38
Transcripts
NM_001142964.1 XM_011530373.2 XM_017028939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
397157 c.109C>T p.Q37* Chr.22: 41690736-41690736 on Build GRCh38
5571607 c.157G>A p.V53I Chr.22: 41690784-41690784 on Build GRCh38
7275131 c.13C>T p.L5L Chr.22: 41690640-41690640 on Build GRCh38
4467809 c.150C>G p.V50V Chr.22: 41690777-41690777 on Build GRCh38
7312188 c.4T>G p.L2V Chr.22: 41690631-41690631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030521270 Chr.22: 41690763-41690763 on Build GRCh38
rs1037684845 Chr.22: 41690688-41690688 on Build GRCh38
rs557093487 Chr.22: 41690761-41690761 on Build GRCh38
rs968568405 Chr.22: 41690788-41690788 on Build GRCh38
rs557164693 Chr.22: 41690767-41690767 on Build GRCh38
rs764167396 Chr.22: 41690850-41690850 on Build GRCh38
rs907947524 Chr.22: 41690679-41690679 on Build GRCh38
rs376324567 Chr.22: 41690840-41690840 on Build GRCh38
rs939180115 Chr.22: 41690629-41690631 on Build GRCh38
rs188171323 Chr.22: 41690664-41690664 on Build GRCh38
rs992937206 Chr.22: 41690666-41690666 on Build GRCh38
rs373288611 Chr.22: 41690787-41690787 on Build GRCh38
rs773755523 Chr.22: 41690724-41690724 on Build GRCh38
rs773870270 Chr.22: 41690852-41690852 on Build GRCh38
rs753442233 Chr.22: 41690740-41690740 on Build GRCh38
rs774116654 Chr.22: 41690687-41690687 on Build GRCh38
rs940688074 Chr.22: 41690684-41690684 on Build GRCh38
rs550026549 Chr.22: 41690790-41690790 on Build GRCh38
rs185614924 Chr.22: 41690765-41690765 on Build GRCh38
rs757175549 Chr.22: 41690749-41690749 on Build GRCh38
rs562336145 Chr.22: 41690848-41690848 on Build GRCh38
rs942277225 Chr.22: 41690730-41690730 on Build GRCh38
rs780269977 Chr.22: 41690781-41690781 on Build GRCh38
rs890666817 Chr.22: 41690775-41690775 on Build GRCh38
rs763484672 Chr.22: 41690633-41690633 on Build GRCh38
rs748294219 Chr.22: 41690808-41690808 on Build GRCh38
rs770418550 Chr.22: 41690844-41690844 on Build GRCh38
rs1007383378 Chr.22: 41690766-41690766 on Build GRCh38
rs193225366 Chr.22: 41690750-41690750 on Build GRCh38
rs760894733 Chr.22: 41690690-41690690 on Build GRCh38
rs927853528 Chr.22: 41690628-41690628 on Build GRCh38
rs892186446 Chr.22: 41690819-41690819 on Build GRCh38
rs543641142 Chr.22: 41690807-41690807 on Build GRCh38

View Full Product Details