Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7426292-7426527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf74
Entrez Gene ID
201243
Gene Name
chromosome 17 open reading frame 74
Gene Aliases
-
UniGene
Hs.380704
Gene Chromosome Location
Chr.17: 7424874-7427568 on Build GRCh38
Transcripts
NM_175734.4 XM_006721471.3 XM_011523722.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3522091 c.321C>T p.F107F Chr.17: 7426312-7426312 on Build GRCh38
1386291 c.378C>T p.R126R Chr.17: 7426369-7426369 on Build GRCh38
7337867 c.385C>T p.R129C Chr.17: 7426376-7426376 on Build GRCh38
6442469 c.323C>T p.S108F Chr.17: 7426314-7426314 on Build GRCh38
3522094 c.376C>T p.R126C Chr.17: 7426367-7426367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776303840 Chr.17: 7426379-7426379 on Build GRCh38
rs372621817 Chr.17: 7426426-7426426 on Build GRCh38
rs780752200 Chr.17: 7426505-7426505 on Build GRCh38
rs763001095 Chr.17: 7426355-7426355 on Build GRCh38
rs373306241 Chr.17: 7426394-7426394 on Build GRCh38
rs766653067 Chr.17: 7426481-7426481 on Build GRCh38
rs375205850 Chr.17: 7426386-7426386 on Build GRCh38
rs746634106 Chr.17: 7426333-7426333 on Build GRCh38
rs746359564 Chr.17: 7426371-7426371 on Build GRCh38
rs761990435 Chr.17: 7426467-7426467 on Build GRCh38
rs770872157 Chr.17: 7426429-7426429 on Build GRCh38
rs746575644 Chr.17: 7426376-7426376 on Build GRCh38
rs777533713 Chr.17: 7426380-7426381 on Build GRCh38
rs768070648 Chr.17: 7426378-7426378 on Build GRCh38
rs747792604 Chr.17: 7426425-7426425 on Build GRCh38
rs767671028 Chr.17: 7426375-7426383 on Build GRCh38
rs757683230 Chr.17: 7426312-7426312 on Build GRCh38
rs765489622 Chr.17: 7426471-7426471 on Build GRCh38
rs375550083 Chr.17: 7426436-7426436 on Build GRCh38
rs201970262 Chr.17: 7426332-7426332 on Build GRCh38
rs761616719 Chr.17: 7426346-7426346 on Build GRCh38
rs751636242 Chr.17: 7426367-7426375 on Build GRCh38
rs532822142 Chr.17: 7426421-7426421 on Build GRCh38
rs373020435 Chr.17: 7426385-7426385 on Build GRCh38
rs185338080 Chr.17: 7426373-7426373 on Build GRCh38
rs780557961 Chr.17: 7426410-7426410 on Build GRCh38
rs759404351 Chr.17: 7426447-7426447 on Build GRCh38
rs368209752 Chr.17: 7426499-7426499 on Build GRCh38
rs771033627 Chr.17: 7426387-7426387 on Build GRCh38
rs768767420 Chr.17: 7426458-7426458 on Build GRCh38
rs113604978 Chr.17: 7426383-7426383 on Build GRCh38
rs76686847 Chr.17: 7426389-7426389 on Build GRCh38
rs754448060 Chr.17: 7426417-7426417 on Build GRCh38
rs754964733 Chr.17: 7426370-7426371 on Build GRCh38
rs376609224 Chr.17: 7426395-7426395 on Build GRCh38
rs369616873 Chr.17: 7426388-7426388 on Build GRCh38
rs773219913 Chr.17: 7426353-7426353 on Build GRCh38
rs749118101 Chr.17: 7426381-7426383 on Build GRCh38
rs562184697 Chr.17: 7426412-7426412 on Build GRCh38
rs769457408 Chr.17: 7426362-7426363 on Build GRCh38
rs758969340 Chr.17: 7426372-7426372 on Build GRCh38
rs1043510722 Chr.17: 7426375-7426375 on Build GRCh38
rs762078413 Chr.17: 7426396-7426396 on Build GRCh38
rs193081398 Chr.17: 7426365-7426365 on Build GRCh38
rs774305175 Chr.17: 7426442-7426442 on Build GRCh38
rs752050541 Chr.17: 7426491-7426491 on Build GRCh38
rs765159571 Chr.17: 7426351-7426351 on Build GRCh38
rs375353507 Chr.17: 7426336-7426336 on Build GRCh38
rs1051007599 Chr.17: 7426384-7426384 on Build GRCh38
rs201512359 Chr.17: 7426370-7426370 on Build GRCh38
rs760407519 Chr.17: 7426355-7426355 on Build GRCh38
rs867783282 Chr.17: 7426377-7426377 on Build GRCh38
rs752854277 Chr.17: 7426377-7426378 on Build GRCh38
rs368240930 Chr.17: 7426475-7426475 on Build GRCh38
rs199717685 Chr.17: 7426427-7426427 on Build GRCh38
rs370916463 Chr.17: 7426498-7426498 on Build GRCh38
rs200196310 Chr.17: 7426401-7426401 on Build GRCh38
rs767695073 Chr.17: 7426356-7426356 on Build GRCh38
rs141473851 Chr.17: 7426466-7426466 on Build GRCh38
rs13290 Chr.17: 7426313-7426313 on Build GRCh38
rs756344162 Chr.17: 7426378-7426383 on Build GRCh38
rs763551080 Chr.17: 7426367-7426369 on Build GRCh38
rs780263561 Chr.17: 7426476-7426476 on Build GRCh38
rs757551229 Chr.17: 7426367-7426367 on Build GRCh38
rs752890609 Chr.17: 7426357-7426357 on Build GRCh38
rs995520094 Chr.17: 7426414-7426414 on Build GRCh38
rs367658611 Chr.17: 7426364-7426364 on Build GRCh38
rs763259396 Chr.17: 7426478-7426478 on Build GRCh38
rs750823327 Chr.17: 7426398-7426398 on Build GRCh38
rs765579933 Chr.17: 7426397-7426397 on Build GRCh38
rs773768189 Chr.17: 7426366-7426367 on Build GRCh38
rs747765542 Chr.17: 7426380-7426380 on Build GRCh38

View Full Product Details