Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100143032-100143301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT13
Entrez Gene ID
54482
Gene Name
tRNA methyltransferase 13 homolog
Gene Aliases
CCDC76
UniGene
Hs.729095
Gene Chromosome Location
Chr.1: 100133080-100150498 on Build GRCh38
Transcripts
NM_019083.2 NR_135078.1 XM_005270945.3 XM_005270946.2 XM_017001504.1 XM_017001505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
458759 c.673G>C p.D225H Chr.1: 100143140-100143140 on Build GRCh38
7318885 c.698C>T p.S233L Chr.1: 100143165-100143165 on Build GRCh38
4019797 c.711A>C p.R237S Chr.1: 100143178-100143178 on Build GRCh38
6384272 c.677G>A p.G226E Chr.1: 100143144-100143144 on Build GRCh38
5429036 c.694A>C p.N232H Chr.1: 100143161-100143161 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745356016 Chr.1: 100143200-100143200 on Build GRCh38
rs764024446 Chr.1: 100143235-100143235 on Build GRCh38
rs769363843 Chr.1: 100143214-100143214 on Build GRCh38
rs571400532 Chr.1: 100143247-100143247 on Build GRCh38
rs778222974 Chr.1: 100143155-100143155 on Build GRCh38
rs376859233 Chr.1: 100143197-100143197 on Build GRCh38
rs376500849 Chr.1: 100143182-100143182 on Build GRCh38
rs369176279 Chr.1: 100143239-100143239 on Build GRCh38
rs758820755 Chr.1: 100143152-100143152 on Build GRCh38
rs200317284 Chr.1: 100143237-100143237 on Build GRCh38
rs755061030 Chr.1: 100143127-100143127 on Build GRCh38
rs756318383 Chr.1: 100143101-100143101 on Build GRCh38
rs951935168 Chr.1: 100143222-100143222 on Build GRCh38
rs748121526 Chr.1: 100143253-100143254 on Build GRCh38
rs762821844 Chr.1: 100143233-100143233 on Build GRCh38
rs779530111 Chr.1: 100143131-100143131 on Build GRCh38
rs775396068 Chr.1: 100143224-100143224 on Build GRCh38
rs759782583 Chr.1: 100143240-100143240 on Build GRCh38
rs777022281 Chr.1: 100143141-100143141 on Build GRCh38
rs147022991 Chr.1: 100143153-100143153 on Build GRCh38
rs79184268 Chr.1: 100143186-100143186 on Build GRCh38
rs1034274394 Chr.1: 100143173-100143173 on Build GRCh38
rs766602051 Chr.1: 100143253-100143253 on Build GRCh38
rs774156521 Chr.1: 100143236-100143236 on Build GRCh38
rs747336826 Chr.1: 100143167-100143167 on Build GRCh38
rs182984724 Chr.1: 100143118-100143118 on Build GRCh38
rs770573316 Chr.1: 100143172-100143172 on Build GRCh38
rs748729682 Chr.1: 100143142-100143142 on Build GRCh38

View Full Product Details