Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22065042-22065303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMAS
Entrez Gene ID
55907
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
CSS
UniGene
Hs.311346
Gene Chromosome Location
Chr.12: 22046174-22065674 on Build GRCh38
Transcripts
NM_018686.5 NR_135117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201480 c.1117A>C p.N373H Chr.12: 22065123-22065123 on Build GRCh38
342133 c.1162G>T p.G388C Chr.12: 22065168-22065168 on Build GRCh38
1201481 c.1276G>A p.E426K Chr.12: 22065282-22065282 on Build GRCh38
108899 c.1161T>G p.S387R Chr.12: 22065167-22065167 on Build GRCh38
4040889 c.1164C>T p.G388G Chr.12: 22065170-22065170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762525283 Chr.12: 22065192-22065192 on Build GRCh38
rs759276539 Chr.12: 22065166-22065166 on Build GRCh38
rs748593061 Chr.12: 22065101-22065101 on Build GRCh38
rs988006872 Chr.12: 22065070-22065070 on Build GRCh38
rs765385723 Chr.12: 22065170-22065170 on Build GRCh38
rs756382670 Chr.12: 22065229-22065229 on Build GRCh38
rs139408586 Chr.12: 22065167-22065167 on Build GRCh38
rs775211156 Chr.12: 22065180-22065180 on Build GRCh38
rs769216725 Chr.12: 22065082-22065082 on Build GRCh38
rs191492408 Chr.12: 22065237-22065237 on Build GRCh38
rs1019693008 Chr.12: 22065172-22065172 on Build GRCh38
rs1036764947 Chr.12: 22065185-22065185 on Build GRCh38
rs558349772 Chr.12: 22065221-22065221 on Build GRCh38
rs544039851 Chr.12: 22065227-22065227 on Build GRCh38
rs772573747 Chr.12: 22065269-22065269 on Build GRCh38
rs780117957 Chr.12: 22065075-22065075 on Build GRCh38
rs200885404 Chr.12: 22065111-22065111 on Build GRCh38
rs1008265621 Chr.12: 22065091-22065091 on Build GRCh38
rs764144464 Chr.12: 22065193-22065193 on Build GRCh38
rs749356471 Chr.12: 22065076-22065076 on Build GRCh38
rs751476700 Chr.12: 22065198-22065198 on Build GRCh38
rs140709416 Chr.12: 22065226-22065226 on Build GRCh38
rs943765168 Chr.12: 22065184-22065184 on Build GRCh38
rs779000961 Chr.12: 22065248-22065248 on Build GRCh38
rs375175648 Chr.12: 22065204-22065204 on Build GRCh38
rs748608460 Chr.12: 22065249-22065249 on Build GRCh38
rs779114414 Chr.12: 22065073-22065074 on Build GRCh38
rs146887115 Chr.12: 22065215-22065215 on Build GRCh38
rs755025797 Chr.12: 22065238-22065238 on Build GRCh38
rs774687479 Chr.12: 22065096-22065096 on Build GRCh38
rs773560587 Chr.12: 22065164-22065164 on Build GRCh38
rs750601553 Chr.12: 22065092-22065093 on Build GRCh38

View Full Product Details