Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137264230-137264455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFB
Entrez Gene ID
25920
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1, NELF-B
UniGene
Hs.521809
Gene Chromosome Location
Chr.9: 137255300-137273548 on Build GRCh38
Transcripts
NM_015456.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
136760 c.803C>T p.A268V Chr.9: 137264264-137264264 on Build GRCh38
4597263 c.844G>C p.E282Q Chr.9: 137264305-137264305 on Build GRCh38
5472718 c.868G>A p.V290I Chr.9: 137264329-137264329 on Build GRCh38
3996470 c.849G>A p.L283L Chr.9: 137264310-137264310 on Build GRCh38
4922695 c.896+1G>T p.? Chr.9: 137264358-137264358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027673038 Chr.9: 137264400-137264400 on Build GRCh38
rs766552368 Chr.9: 137264250-137264250 on Build GRCh38
rs952262825 Chr.9: 137264415-137264415 on Build GRCh38
rs372861276 Chr.9: 137264396-137264396 on Build GRCh38
rs371864566 Chr.9: 137264302-137264302 on Build GRCh38
rs976282924 Chr.9: 137264433-137264433 on Build GRCh38
rs758978015 Chr.9: 137264300-137264300 on Build GRCh38
rs776775232 Chr.9: 137264348-137264348 on Build GRCh38
rs372044179 Chr.9: 137264271-137264271 on Build GRCh38
rs376785409 Chr.9: 137264340-137264340 on Build GRCh38
rs765371667 Chr.9: 137264289-137264289 on Build GRCh38
rs371900282 Chr.9: 137264366-137264366 on Build GRCh38
rs139131908 Chr.9: 137264283-137264283 on Build GRCh38
rs757737702 Chr.9: 137264303-137264303 on Build GRCh38
rs563679522 Chr.9: 137264404-137264404 on Build GRCh38
rs182608046 Chr.9: 137264301-137264301 on Build GRCh38
rs187618743 Chr.9: 137264432-137264432 on Build GRCh38
rs375835312 Chr.9: 137264273-137264273 on Build GRCh38
rs867700400 Chr.9: 137264262-137264262 on Build GRCh38
rs966643705 Chr.9: 137264425-137264425 on Build GRCh38
rs755523471 Chr.9: 137264328-137264328 on Build GRCh38
rs1017767468 Chr.9: 137264424-137264424 on Build GRCh38
rs1014334031 Chr.9: 137264365-137264365 on Build GRCh38
rs199996299 Chr.9: 137264290-137264290 on Build GRCh38
rs146597494 Chr.9: 137264322-137264322 on Build GRCh38
rs762993559 Chr.9: 137264369-137264369 on Build GRCh38
rs369797870 Chr.9: 137264344-137264344 on Build GRCh38
rs780992274 Chr.9: 137264310-137264310 on Build GRCh38
rs768612199 Chr.9: 137264333-137264333 on Build GRCh38
rs374702356 Chr.9: 137264367-137264367 on Build GRCh38
rs762259538 Chr.9: 137264401-137264401 on Build GRCh38
rs374696743 Chr.9: 137264375-137264375 on Build GRCh38
rs749128632 Chr.9: 137264332-137264332 on Build GRCh38
rs779622658 Chr.9: 137264329-137264329 on Build GRCh38
rs768010506 Chr.9: 137264408-137264408 on Build GRCh38
rs922282419 Chr.9: 137264279-137264279 on Build GRCh38
rs752180047 Chr.9: 137264398-137264398 on Build GRCh38
rs993476405 Chr.9: 137264390-137264390 on Build GRCh38
rs369375421 Chr.9: 137264374-137264374 on Build GRCh38
rs13295537 Chr.9: 137264419-137264419 on Build GRCh38
rs747928505 Chr.9: 137264343-137264343 on Build GRCh38

View Full Product Details